Skip to Content
Merck
All Photos(2)

Key Documents

89432

Supelco

Glyphosate

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

N-(Phosphonomethyl)glycine, Glyphosate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$94.90
250 MG
$135.20
500 MG
$254.80
1 G
$369.20
2.5 G
$725.40
5 G
$1,072.50
10 G
$1,591.20

Vendor SKU: EN300-22719-100MG

$94.90


Availability
Ships in 7 days - from Aldrich Partner.

Request a Bulk Order

Select a Size

Change View
100 MG
$94.90
250 MG
$135.20
500 MG
$254.80
1 G
$369.20
2.5 G
$725.40
5 G
$1,072.50
10 G
$1,591.20

About This Item

Linear Formula:
(HO)2P(O)CH2NHCH2CO2H
CAS Number:
Molecular Weight:
169.07
Beilstein:
2045054
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Vendor SKU: EN300-22719-100MG

$94.90


Availability
Ships in 7 days - from Aldrich Partner.

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

gas chromatography (GC): suitable

mp

230 °C (dec.) (lit.)

format

neat

SMILES string

OC(=O)CNCP(O)(O)=O

InChI

1S/C3H8NO5P/c5-3(6)1-4-2-10(7,8)9/h4H,1-2H2,(H,5,6)(H2,7,8,9)

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chelsea M Smith et al.
Aquatic toxicology (Amsterdam, Netherlands), 210, 215-226 (2019-03-16)
Roundup and other glyphosate-based herbicides are the most commonly used herbicides in the world, yet their effects on developing fish embryos are not clearly understood. The present study, therefore, examined developmental teratogenic effects and adult-onset reproductive effects of exposure to
Thu Ha Pham et al.
Toxicological sciences : an official journal of the Society of Toxicology, 169(1), 260-271 (2019-02-21)
Glyphosate is the most widely used herbicide in the world. Several studies have investigated the effects of glyphosate and glyphosate-based herbicides (GBHs) on male reproduction, but there is still little and conflicting evidence for its toxicity. In this study, we
Anna Barbasz et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 55(8), 719-725 (2020-06-17)
The industrialization of the agricultural sector has significantly increased the use of chemicals such as pesticides. Therefore, exposure to them is unavoidable, which makes it necessary to assess their safety for humans at actual exposure doses. This paper aims to

Articles

A precise U/HPLC-MS/MS method with QuEChERS extraction for the analysis of glyphosate, glufosinate, and AMPA in green tea.

A precise U/HPLC-MS/MS method with QuEChERS extraction for the analysis of glyphosate, glufosinate, and AMPA in green tea.

A precise U/HPLC-MS/MS method with QuEChERS extraction for the analysis of glyphosate, glufosinate, and AMPA in green tea.

A precise U/HPLC-MS/MS method with QuEChERS extraction for the analysis of glyphosate, glufosinate, and AMPA in green tea.

See All

Protocols

EPA Method 547 outlines the analysis of glyphosate in drinking water by direct aqueous injection HPLC, post column derivatization, and fluorescence detection

Protocol for LC/MS Analysis of Glyphosate and Metabolites on apHera™ NH2, 2 mm I.D. Column

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service