Skip to Content
Merck
All Photos(2)

Key Documents

05164

Supelco

(Aminomethyl)phosphonic acid

PESTANAL®, analytical standard

Synonym(s):

AMPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2P(O)(OH)2
CAS Number:
Molecular Weight:
111.04
Beilstein:
1701219
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (TLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤3.0% water (calc. from elemental analysis)

application(s)

agriculture
environmental

format

neat

SMILES string

NCP(O)(O)=O

InChI

1S/CH6NO3P/c2-1-6(3,4)5/h1-2H2,(H2,3,4,5)

InChI key

MGRVRXRGTBOSHW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seonil Kim et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(10), 3122-3127 (2015-02-26)
Gene knockout (KO) does not always result in phenotypic changes, possibly due to mechanisms of functional compensation. We have studied mice lacking cGMP-dependent kinase II (cGKII), which phosphorylates GluA1, a subunit of AMPA receptors (AMPARs), and promotes hippocampal long-term potentiation
Philip Jean-Richard Dit Bressel et al.
PloS one, 9(11), e111699-e111699 (2014-11-05)
The lateral habenula (LHb) is a small epithalamic structure that projects via the fasciculus retroflexus to the midbrain. The LHb is known to modulate midbrain dopamine (DA) neurons, including inhibition of ventral tegmental area (VTA) neurons via glutamatergic excitation of
Martin Puskarjov et al.
Neuropharmacology, 88, 103-109 (2014-09-18)
A robust increase in the functional expression of the neuronal K-Cl cotransporter KCC2 during CNS development is necessary for the emergence of hyperpolarizing ionotropic GABAergic transmission. BDNF-TrkB signaling has been implicated in the developmental up-regulation of KCC2 and, in mature
Mary L Huff et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 40(4), 861-873 (2014-09-27)
Recent findings suggest that the mesolimbic dopamine neurons, known to promote cocaine-seeking behavior, are strongly inhibited by a newly characterized region of the midbrain known as the rostromedial tegmental nucleus (RMTg). The RMTg appears to be involved in generating reward-prediction
W Zhou et al.
European psychiatry : the journal of the Association of European Psychiatrists, 29(7), 419-423 (2013-12-11)
Ketamine exerts fast acting, robust, and lasting antidepressant effects in a sub-anesthetic dose, however, the underlying mechanisms are still not fully elucidated. Recent studies have suggested that ketamine's antidepressant effects are probably attributed to the activation of α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid (AMPA)

Articles

A precise U/HPLC-MS/MS method with QuEChERS extraction for the analysis of glyphosate, glufosinate, and AMPA in green tea.

A precise U/HPLC-MS/MS method with QuEChERS extraction for the analysis of glyphosate, glufosinate, and AMPA in green tea.

Glyphosate and related compounds are measured in oatmeal and infant cereal using ion-exchange polymer-based particles for HPLC and SPE. Low level detection was obtained.

Glyphosate and related compounds are measured in oatmeal and infant cereal using ion-exchange polymer-based particles for HPLC and SPE. Low level detection was obtained.

See All

Protocols

EPA Method 547 outlines the analysis of glyphosate in drinking water by direct aqueous injection HPLC, post column derivatization, and fluorescence detection

Protocol for LC/MS Analysis of Glyphosate and Metabolites on apHera™ NH2, 2 mm I.D. Column

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service