Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Glyphosate
Synonym(s):
N
-(Phosphonomethyl)glycine, Glyphosate
Linear Formula:
(HO)
2
P(O)CH
2
NHCH
2
CO
2
H
CAS Number:
1071-83-6
Molecular Weight:
169.07
EC Number:
213-997-4
Products
Product Category
pesticides (2)
Brand
Supelco (2)
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
analytical standard (1)
certified reference material (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
45521
Glyphosate,
PESTANAL
®
, analytical standard
Expand
View Pricing
89432
Glyphosate,
certified reference material,
Trace
CERT
®
, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Expand
View Pricing