Skip to Content
Merck
All Photos(1)

Key Documents

86140

Sigma-Aldrich

Docusate sodium salt

purum, ≥96.0% (TLC)

Synonym(s):

AOT, Bis(2-ethylhexyl) sulfosuccinate sodium salt, Sodium bis(2-ethylhexyl) sulfosuccinate, Sulfobutanedioic acid bis(2-ethylhexyl ester) sodium salt, Sulfosuccinic acid bis(2-ethylhexyl) ester sodium salt, ‘Dioctyl’ sulfosuccinate sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H37NaO7S
CAS Number:
Molecular Weight:
444.56
Beilstein:
4117588
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥96.0% (TLC)

form

solid

impurities

≤2% water

functional group

ester
sulfonic acid

SMILES string

[Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O

InChI

1S/C20H38O7S.Na/c1-5-9-11-16(7-3)14-26-19(21)13-18(28(23,24)25)20(22)27-15-17(8-4)12-10-6-2;/h16-18H,5-15H2,1-4H3,(H,23,24,25);/q;+1/p-1

InChI key

APSBXTVYXVQYAB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Docusate sodium salt is an emollient which can combine with the active pharmaceutical ingredients (APIs) such as ranitidine hydrochloride and lidocaine hydrochloride to form hydrophobic ionic liquids (IL-APIs)).

Application

Docusate sodium salt can be used as a surfactant in the synthesis of magnetic nanoparticles by co-precipitation method.
Forms reverse micelles in hydrocarbon solvents; Suitable for the solubilization of the major myelin transmembrane proteolipid

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of multiple-layer polymeric particles for targeted and controlled drug delivery
Koppolu B, et al.
Nanomedicine: Nanotechnology, Biology, and Medicine, 6(2), 355-361 (2010)
The third evolution of ionic liquids: active pharmaceutical ingredients
Hough WL, et al.
New. J. Chem., 31(8), 1429-1436 (2007)
In vitro evaluation of novel polymer-coated magnetic nanoparticles for controlled drug delivery
Rahimi M, et al.
Nanomedicine: Nanotechnology, Biology, and Medicine, 6(5), 672-680 (2010)
Johnson Mathew et al.
Journal of chromatography. A, 1231, 46-51 (2012-03-01)
Dioctyl sulfosuccinate (DOSS) was a major component of the dispersants most used in the 2010 Deepwater Horizon Oil Spill incident response. This analytical method quantifies salt water DOSS concentrations to a reporting limit of 20 μg/L, which was below the
Sonali V Kapse et al.
International journal of pharmaceutics, 429(1-2), 104-112 (2012-03-15)
We disclose a self nanoprecipitating preconcentrate (SNP) of tamoxifen citrate (TMX), which forms TMX loaded polymeric nanoparticles, on dilution with aqueous media. SNP comprised TMX, polymer (Kollidon SR) and surfactant/s dissolved in a pharmaceutically acceptable vehicle. Binary surfactant mixtures of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service