Skip to Content
Merck
All Photos(1)

Key Documents

80010

Sigma-Aldrich

Phthalic acid

puriss. p.a., ≥99.5% (T)

Synonym(s):

1,2-Benzenedicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(CO2H)2
CAS Number:
Molecular Weight:
166.13
Beilstein:
608199
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

grade

puriss. p.a.

Quality Level

Assay

≥99.5% (T)

form

powder or crystals

ign. residue

≤0.02% (as SO4)

mp

210-211 °C (dec.) (lit.)

solubility

water: soluble(lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

functional group

carboxylic acid

SMILES string

OC(C1=C(C(O)=O)C=CC=C1)=O

InChI

1S/C8H6O4/c9-7(10)5-3-1-2-4-6(5)8(11)12/h1-4H,(H,9,10)(H,11,12)

InChI key

XNGIFLGASWRNHJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Phthalic acid (1,2-benzenedicarboxylic acid) is an aromatic dicarboxylic acid. On heating, it undergoes dehydration to afford cyclic anhydride. It can be synthesized from phthalic anhydride.[1]

Application

Phthalic acid may be used in the synthesis of benzoic acid.[1] It may be used to constitute the mobile phase during hydrophilic interaction liquid chromatography (HILIC) coupled with electrospray ionization tandem mass spectrometric (ESI-MS/MS) analysis of free amino acids (AAs) in human thyroid tissues.[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

334.4 °F

Flash Point(C)

168 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wanshu Qi et al.
Analytica chimica acta, 870, 75-82 (2015-03-31)
In this work, 0.08 mmol L(-1) of phthalic acid was introduced as a mobile phase additive to quantify free amino acids (AAs) by hydrophilic interaction liquid chromatography (HILIC) coupled with electrospray ionization tandem mass spectrometry (ESI-MS/MS). The addition of phthalic
Paula J Lapinskas et al.
Toxicology, 207(1), 149-163 (2004-12-14)
Phthalate esters belong to a large class of compounds known as peroxisome proliferators (PP). PP include chemicals that activate different subtypes of the peroxisome proliferator-activated receptor (PPAR) family. The ability of phthalate esters and their metabolites to activate responses through
Russ Hauser
Seminars in reproductive medicine, 24(3), 156-167 (2006-06-29)
Scientific and public concern about the potential risk of environmental chemicals to male reproductive health has been heightened by reports of downward trends in semen quality, as well as increased rates of developmental urogenital tract anomalies and testicular cancer. Of
Christopher H Hurst et al.
Toxicological sciences : an official journal of the Society of Toxicology, 74(2), 297-308 (2003-06-14)
Phthalate esters are widely used as plasticizers in the manufacture of products made of polyvinyl chloride. Mono-(2-ethylhexyl)-phthalate (MEHP) induces rodent hepatocarcinogenesis by a mechanism that involves activation of the nuclear transcription factor peroxisome proliferator-activated receptor-alpha (PPARalpha). MEHP also activates PPAR-gamma
Eagleson M.
Concise Encyclopedia Chemistry, 832-832 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service