Skip to Content
Merck
All Photos(2)

Key Documents

84679

Supelco

Naphthalene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8
CAS Number:
Molecular Weight:
128.17
Beilstein:
1421310
EC Number:
MDL number:
UNSPSC Code:
20121904
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.4 (vs air)

vapor pressure

0.03 mmHg ( 25 °C)

Assay

≥99.7% (GC)

autoignition temp.

978 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

5.9 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

218 °C (lit.)

mp

79.5-81.0 °C
80-82 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

c1ccc2ccccc2c1

InChI

1S/C10H8/c1-2-6-10-8-4-3-7-9(10)5-1/h1-8H

InChI key

UFWIBTONFRDIAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Naphthalene is widely used as an intermediate in the chemical and plastics industry. It is commonly found in petroleum, coal, mothballs, cigarette smoke, biomass burning, incomplete combustion, etc.

Application

Naphthalene is used as a calibration standard for the real-time quantification with high specificity for naphthalene vapor and other HOVs in industrial environment using NaDos personal exposure monitor (PEM) device.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

173.3 °F - closed cup

Flash Point(C)

78.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Naphthalene DNA adduct formation and tolerance in the lung
Buchholz AB, et al.
Nucl. Instrum. Methods Phys. Res. Sect. B, 438, 119-123 (2019)
NaDos: A real-time, wearable, personal exposure monitor for hazardous organic vapors
Reid M, et al.
Sensors and Actuators B, Chemical, 255, 2996-3003 (2018)
Determination of naphthalene metabolites in human urine by liquid chromatography?mass spectrometry with electrospray ionization
Andreoli R, et al.
Journal of Chromatography A, 847(1-2), 9-17 (1999)
Qiaozhen Liu et al.
Nature genetics, 51(4), 728-738 (2019-02-20)
Characterizing the stem cells responsible for lung repair and regeneration is important for the treatment of pulmonary diseases. Recently, a unique cell population located at the bronchioalveolar-duct junctions has been proposed to comprise endogenous stem cells for lung regeneration. However
Martha L Bustos et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 23(3), 561-569 (2014-11-21)
The contribution of bone marrow cells (BMC) in lung repair is controversial. We previously reported a subpopulation of BMC that express Clara cell secretory protein (CCSP). To determine the contribution of endogenous CCSP(+) BMC to airway regeneration, we performed bone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service