Skip to Content
Merck
All Photos(5)

Key Documents

A88255

Sigma-Aldrich

p-Anisidine

99%

Synonym(s):

4-Aminoanisole, 4-Methoxyaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4NH2
CAS Number:
Molecular Weight:
123.15
Beilstein:
471556
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

autoignition temp.

959 °F

bp

240-243 °C (lit.)

mp

56-59 °C (lit.)

SMILES string

COc1ccc(N)cc1

InChI

1S/C7H9NO/c1-9-7-4-2-6(8)3-5-7/h2-5H,8H2,1H3

InChI key

BHAAPTBBJKJZER-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

p-Anisidine can undergo aerobic oxidation in the presence of gold catalysts to form aromatic azo compounds.
p-Anisidine can be used:
  • In the diastereoselective and enantioselective synthesis of CF3-substituted azoridines catalyzed by a chiral Bronsted acid.
  • To prepare 4-organoselenium-quinolines through multi-component Povarov reaction with ethyl glyoxylate and ethynyl(phenyl)selane, catalyzed by Yb(OTf)3.
  • As a starting material to synthesize 3-fluoro-6-methoxyquinoline in two steps.,
  • To prepare N-PMP protected α-aminopropargylphosphonates by reacting with terminal alkynes and diethyl formylphosphonate hydrate using silver(I) triflate as a catalyst.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Carc. 1B - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

251.6 °F - closed cup

Flash Point(C)

122 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noël Tenyang et al.
Food chemistry, 221, 1308-1316 (2016-12-17)
The aim of this study was to determine the effect of boiling and roasting on the proximate, lipid oxidation, fatty acid profile and mineral content of two sesame seeds varieties. The proximate composition was significantly affected (P<0.05) during treatments. The
Ytterbium (III)-catalyzed three-component reactions: synthesis of 4-organoselenium-quinolines
de Oliveira IM
New. J. Chem., 41(18), 9884-9888 (2017)
Dima A Sabbah et al.
Medicinal chemistry (Shariqah (United Arab Emirates)), 14(7), 695-708 (2018-04-14)
Phosphoinositide 3-kinase α (PI3Kα) is an attractive target for anticancer drug design. Target compounds were designed to probe the significance of alcohol and imine moieties tailored on a benzoin scaffold to better understand the structure activity relation (SAR) and improve
A Facile Two-Step Synthesis of 3-Fluoro-6-methoxyquinoline
Li B, et al.
Organic Process Research & Development, 12(6), 1273-1275 (2008)
Silver (I) triflate-catalyzed direct synthesis of N-PMP protected α-aminopropargylphosphonates from terminal alkynes
Dodda R and Zhao C
Organic Letters, 9(1), 165-167 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service