Skip to Content
Merck
All Photos(2)

Key Documents

76137

Supelco

Stearic acid

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

1-Heptadecanecarboxylic acid, C18:0, Cetylacetic acid, NSC 25956, NSC 261168, Octadecanoic acid, Stearophanic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16COOH
CAS Number:
Molecular Weight:
284.48
Beilstein:
608585
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
E Number:
E570
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

vapor pressure

1 mmHg ( 173.7 °C)

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

361 °C (lit.)

mp

67-72 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)

InChI key

QIQXTHQIDYTFRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palle J Pedersen et al.
Journal of medicinal chemistry, 53(9), 3782-3792 (2010-04-22)
The design of retinoid phospholipid prodrugs is described based on molecular dynamics simulations and cytotoxicity studies of synthetic retinoid esters. The prodrugs are degradable by secretory phospholipase A(2) IIA and have potential in liposomal drug delivery targeting tumors. We have
Ulla Hoppu et al.
Journal of pediatric gastroenterology and nutrition, 41(3), 335-338 (2005-09-01)
Breast milk fatty acids may have immunomodulatory properties related to the development of atopic disease. The aim of this study was to assess the impact of the breast milk fatty acid composition on the development of atopic dermatitis (AD) in
T A Doran et al.
American journal of obstetrics and gynecology, 133(3), 302-307 (1979-02-01)
Lecithin/sphingomyelin (L/S) ratio, creatinine, percentage of lipid-positive cells, palmitic acid, palmitic acid/stearic acid (P/S) ratio, and total cortisol were analyzed as tests for fetal maturity and fetal pulmonary maturity in 164 samples of amniotic fluid from 121 patients. Fifty samples
Qian Wan et al.
Nanoscale, 5(2), 744-752 (2012-12-12)
As a versatile gene vector, minicircle DNA (mcDNA) has a great potential for gene therapy. However, some serious challenges remain, such as to effectively deliver mcDNA into targeted cells/tissues and to non-invasively monitor the delivery of the mcDNA. Superparamagnetic iron
Piret Arukuusk et al.
Biochimica et biophysica acta, 1828(5), 1365-1373 (2013-01-30)
Harnessing of a branched structure is a novel approach in the design of cell-penetrating peptides and it has provided highly efficient transfection reagents for intracellular delivery of nucleic acids. The new stearylated TP10 analogs, NickFects, condense plasmid DNA, splice correcting

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service