Skip to Content
Merck
All Photos(1)

Key Documents

W303518

Sigma-Aldrich

Stearic acid

≥95%, FCC, FG

Synonym(s):

1-Heptadecanecarboxylic acid, C18:0, Cetylacetic acid, NSC 25956, NSC 261168, Octadecanoic acid, Stearophanic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16COOH
CAS Number:
Molecular Weight:
284.48
FEMA Number:
3035
Beilstein:
608585
EC Number:
Council of Europe no.:
15
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.015
NACRES:
NA.21

biological source

palm oil

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012
FCC
FDA 21 CFR 172.210
FDA 21 CFR 172.615
FDA 21 CFR 172.860

vapor pressure

1 mmHg ( 173.7 °C)

Assay

≥95%

form

flakes
powder or solid

bp

361 °C (lit.)

mp

67-72 °C (lit.)

density

0.845 g/cm3

cation traces

As: ≤3 ppm
Cd: ≤1 ppm
Hg: ≤1 ppm
Pb: ≤2 ppm

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

SMILES string

CCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)

InChI key

QIQXTHQIDYTFRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Stearic acid is a saturated fatty acid found in high quantity in animal fat. It is also found in cottonseed oil, pumpkin seed oil, grape seed oil and also in pomegranate seeds. Stearic acid plays an important role as an emulsifier in the cosmetic industry.

Application


  • Effects of glycyrrhiza polysaccharides on growth performance, meat quality, serum parameters and growth/meat quality-related gene expression in broilers.: This study explored the impact of glycyrrhiza polysaccharides on broiler chickens, noting changes in serum levels of stearic acid, which could influence meat quality and overall health. (Li et al., 2024).

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genetics and Genomics of Cotton, 3, 366-366 (2009)
Pumpkin (Cucurbita pepo L.) seed oil as an alternative feedstock for the production of biodiesel in Greece.
Schinas P, et al.
Biomass and Bioenergy, 33(1), 44-49 (2009)
Chemical composition of juice and seeds of pomegranate fruit.
El?Nemr SE, et al.
Molecular Nutrition And Food Research, 34(7),, 601-606 (1990)
Oil content and oil quality properties of some grape seeds.
Baydar NG & Akkurt M.
Turkish Journal of Agriculture and Forestry, 25(3), 163-168 (2001)
Palle J Pedersen et al.
Journal of medicinal chemistry, 53(9), 3782-3792 (2010-04-22)
The design of retinoid phospholipid prodrugs is described based on molecular dynamics simulations and cytotoxicity studies of synthetic retinoid esters. The prodrugs are degradable by secretory phospholipase A(2) IIA and have potential in liposomal drug delivery targeting tumors. We have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service