Skip to Content
Merck
All Photos(2)

Key Documents

F7258

Sigma-Aldrich

Fast Green FCF

certified by the Biological Stain Commission

Synonym(s):

Food green 3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C37H34N2O10S3Na2
CAS Number:
Molecular Weight:
808.85
Colour Index Number:
42053
Beilstein:
5718212
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

certified by the Biological Stain Commission

Quality Level

form

powder

composition

dye content, ≥90%

mp

290 °C (dec.) (lit.)

solubility

H2O: 1 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].CCN(Cc1cccc(c1)S([O-])(=O)=O)c2ccc(cc2)C(=C3/C=CC(\C=C3)=[N+](\CC)Cc4cccc(c4)S([O-])(=O)=O)\c5ccc(O)cc5S([O-])(=O)=O

InChI

1S/C37H36N2O10S3.2Na/c1-3-38(24-26-7-5-9-33(21-26)50(41,42)43)30-15-11-28(12-16-30)37(35-20-19-32(40)23-36(35)52(47,48)49)29-13-17-31(18-14-29)39(4-2)25-27-8-6-10-34(22-27)51(44,45)46;;/h5-23H,3-4,24-25H2,1-2H3,(H3,41,42,43,44,45,46,47,48,49);;/q;2*+1/p-2

InChI key

RZSYLLSAWYUBPE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Fast Green FCF has been used:
  • along with safranin for the staining of nodule sections
  • in the collagen deposit staining procedure
  • for the staining of protein and starch
  • as a dye to detect and inject DNA solution in chicken embryo
Fast Green FCF dye is used for protein staining in IEF, native PAGE and SDS-PAGE. Fast Green staining is linear over a wider range of protein concentrations than Brilliant Blue R. After electrophoresis, fixing the proteins in the gel is recommended for maximum sensitivity. The gel can then be stained for 2 hr with 0.1% Fast Green FCF in either 30% (v/v) ethanol, 10% (v/v) acetic acid or in 7% acetic acid. The gel can be destained with either 30% (v/v) ethanol, 10% (v/v) acetic acid, or 7% acetic acid. The stained gel can be scanned at 625 nm. The detection sensitivity is approx. 30% that with Brilliant Blue R.

Suitability

Certified for the safranin-fast green FCF stain for plant cells; as a substitute for aniline blue in Gomori′s one-step trichrome procedure.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

352.4 °F - closed cup

Flash Point(C)

178 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Combination of in ovo electroporation and time-lapse imaging to study migrational events in chicken embryos.
Masyuk M et al.
Developmental Dynamics, 243, 690-690 (2014)
Micro-morphology of common bean (Phaseolus vulgaris L.) nodules undergoing senescence.
Fernandez-Luque?o F et al.
Acta Physiologiae Plantarum / Polish Academy of Sciences, Committee of Plant Physiology Genetics and Breeding, 30, 545-552 (2008)
Yohann Rautureau et al.
Circulation, 138(16), 1677-1692 (2018-04-21)
Pharmacogenomic studies have shown that ADCY9 genotype determines the effects of the CETP (cholesteryl ester transfer protein) inhibitor dalcetrapib on cardiovascular events and atherosclerosis imaging. The underlying mechanisms responsible for the interactions between ADCY9 and CETP activity have not yet
Involvement of the Protein Network in thein vitroDegradation of Starch from Spaghetti and Lasagne: a Microscopic and Enzymic Study.
Fardet A et al.
Journal of Cereal Science, 27, 133-145 (1998)
Nan Hu et al.
Arthritis research & therapy, 21(1), 167-167 (2019-07-10)
While bone marrow-derived mesenchymal stem cells (BMSC) are established sources for stem cell-based cartilage repair therapy, articular cartilage-derived mesenchymal stem cells from osteoarthritis patients (OA-MSC) are new and potentially attractive candidates. We compared OA-MSC and BMSC in chondrogenic potentials, gene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service