Skip to Content
Merck
All Photos(1)

Key Documents

63908

Supelco

Mesitylene

analytical standard

Synonym(s):

1,3,5-Trimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(CH3)3
CAS Number:
Molecular Weight:
120.19
Beilstein:
906806
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.1 (vs air)

vapor pressure

14 mmHg ( 55 °C)
2.49 mmHg ( 25 °C)

Assay

≥99.8% (GC)

autoignition temp.

1022 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

0.88-6.1 %, 100 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.499 (lit.)
n20/D 1.499

bp

163-166 °C (lit.)

mp

−45 °C (lit.)

density

0.864 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Cc1cc(C)cc(C)c1

InChI

1S/C9H12/c1-7-4-8(2)6-9(3)5-7/h4-6H,1-3H3

InChI key

AUHZEENZYGFFBQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mesitylene, a volatile organic compound (VOC), is a methyl substituted benzene hydrocarbon, widely utilized as a solvent for industrial and research applications.

Application

Mesitylene may be used as an analytical standard for the quantification of the analyte in human biological samples, environmental samples, organic solvent mixtures, and aerosolized jet fuels by gas chromatography (GC) based techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

122.0 °F

Flash Point(C)

50 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

20th European Symposium of Computer Aided Process Engineering: ESCAPE-20
20th European Symposium of Computer Aided Process Engineering (2010)
Katherine D Dietzel et al.
Journal of chromatography. A, 1093(1-2), 11-20 (2005-10-20)
Jet Propellant 8 (JP-8) jet fuel is a kerosene-based fuel containing hundreds of hydrocarbons used by the military in NATO countries. Previous rodent inhalation studies carried out with aerosolized JP-8 never evaluated the exposure chamber atmosphere. For this reason, our
A valence force field for alkyl benzenes toluene, p-xylene, m-xylene mesitylene, and some of their deuterated analogues
La Lau C and Snyder RG
Spectrochimica Acta Part A: Molecular Spectroscopy, 27(10), 2073-2088 (1971)
Solid-phase microextraction (SPME) for rapid field sampling and analysis by gas chromatography-mass spectrometry (GC-MS)
Hook.LG, et al.
TrAC, Trends in Analytical Chemistry, 21(8), 534-543 (2002)
Gas chromatographic analysis of organic solvent mixtures on capillary columns of different polarity
Rastogi SC
Chromatographia, 37(3-4), 211-214 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service