Skip to Content
Merck
All Photos(1)

Key Documents

140864

Sigma-Aldrich

Mesitylene

reagent grade, 97%

Synonym(s):

1,3,5-Trimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(CH3)3
CAS Number:
Molecular Weight:
120.19
Beilstein:
906806
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

reagent grade

Quality Level

vapor density

4.1 (vs air)

vapor pressure

14 mmHg ( 55 °C)
2.49 mmHg ( 25 °C)

Assay

97%

form

liquid

autoignition temp.

1022 °F

expl. lim.

0.88-6.1 %, 100 °F

refractive index

n20/D 1.499 (lit.)

bp

163-166 °C (lit.)

mp

−45 °C (lit.)

density

0.864 g/mL at 25 °C (lit.)

SMILES string

Cc1cc(C)cc(C)c1

InChI

1S/C9H12/c1-7-4-8(2)6-9(3)5-7/h4-6H,1-3H3

InChI key

AUHZEENZYGFFBQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Binders for metals in industrial processes: A study explored the use of hydroxyl-terminated dendrimers with sulfonimide linkers, where mesitylene was employed as a critical solvent in the synthesis process, offering potential in industrial metal binding applications (Abu Sbeih and Al Harahsheh, 2024).
  • Permeation networks for selective solvent filtration: Research demonstrated the use of mesitylene in creating superoleophilic, chemically and mechanically resistant percolation networks of ZIF-7 and ZIF-11 for the selective permeation of oils and chlorinated solvents, indicating its utility in industrial chemical processes (Prasanthi and Datta, 2023).
  • Green synthesis of covalent organic frameworks: A review highlighted the role of mesitylene in the green and facile preparation of covalent organic frameworks, emphasizing its significance in sustainable and advanced applications in chemical engineering (Azadi and Dinari, 2023).

Packaging

Packaged in glass bottles

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

122.0 °F

Flash Point(C)

50 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christoph Sprung et al.
Journal of the American Chemical Society, 137(5), 1916-1928 (2015-01-16)
A detailed and systematic polarized confocal fluorescence microscopy investigation is presented on three batches of large coffin-shaped ZSM-5 crystals (i.e., parent, steamed at 500 °C, and steamed at 700 °C). In total, six laser lines of different wavelength in the
Dalit Rechavi et al.
Journal of the American Chemical Society, 126(25), 7738-7739 (2004-06-24)
Isotope effects for the encapsulation of deuterated versus nondeuterated guests are determined. This system involves the use of social isomers, and the origin of the isotope effect is localized to methyl groups interacting with aromatic rings.
Sanyog Sharma et al.
Organic & biomolecular chemistry, 11(4), 654-661 (2012-12-12)
Two mesitylene based probes, having catechol/phenol units in conjunction to the Schiff base have been synthesized. The probe with a catechol unit can be used to sense and discriminate between F(-) and CN(-) through different colorimetric and fluorimetric responses using
Jacqueline M Veauthier et al.
Inorganic chemistry, 43(4), 1220-1228 (2004-02-18)
New bimetallic mu-oxo diferric complexes of several previously reported calix[4]pyrrole Schiff base macrocycles are described. The synthesis of a new member of this class of macrocycles is also reported; it was prepared via an acid-catalyzed condensation between 1,9-bisformyl-5,5-dipropyldipyrromethane and o-phenylenediamine.
Natalie D Coombs et al.
Dalton transactions (Cambridge, England : 2003), (3)(3), 332-337 (2008-04-17)
Sterically encumbered amido ligands based on a 1,8-diarylcarbazol-9-yl backbone have been investigated as electronically distinct alternatives to the widely-used terphenyl ligand class in the stabilization of low-coordinate metal complexes, and structurally characterized for the first time. While 1,8-diphenylcarbazol-9-yl derivatives are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service