Skip to Content
Merck
All Photos(1)

Key Documents

8.05890

Sigma-Aldrich

Mesitylene

Technipur®, for synthesis

Synonym(s):

1,3,5-Trimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(CH3)3
CAS Number:
Molecular Weight:
120.19
Beilstein:
906806
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.22

grade

for synthesis

Quality Level

vapor density

4.1 (vs air)

vapor pressure

14 mmHg ( 55 °C)
2.49 mmHg ( 25 °C)
2.8 hPa ( 20 °C)

autoignition temp.

1022 °F

expl. lim.

0.88-6.1 %, 100 °F

refractive index

n20/D 1.499 (lit.)

bp

163-166 °C (lit.)

mp

−45 °C (lit.)

density

0.864 g/mL at 25 °C (lit.)

SMILES string

Cc1cc(C)cc(C)c1

InChI

1S/C9H12/c1-7-4-8(2)6-9(3)5-7/h4-6H,1-3H3

InChI key

AUHZEENZYGFFBQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mesitylene is a methyl-substituted benzene hydrocarbon, a specialty solvent used for research and industrial applications. Derivatives of mesitylene are used as colorant precursors to prepare organic dye intermediates such as 2,4,6-trimethylaniline.

Application

  • Mesitylene is used as a:
  • building block to manufacture antioxidants, plasticizers, rubber, adhesives, etc.
  • ligand in organometallic reactions
  • developer for photo patternable silicones
  • precursor to prepare a variety of fine chemicals

Legal Information

Technipur is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

122.0 °F

Flash Point(C)

50 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of catalyst morphology and hydrogen co-feeding on the acid-catalysed transformation of acetone into mesitylene
Quesada J, et al
Catalysis Science & Technology, 10, 1356-1367 (2020)
Comparative study of theoretical ultrasonic velocities of binary liquid mixtures containing quinoline and mesitylene at temperatures T = (303.15,308.15,313.15 and 318.15) K
Babavali SKF, et al.
Karbala International Journal of Modern Science, 1(3), 172-177 (2015)
Metal versus Ligand Reduction in Ln3+ Complexes of a Mesitylene-Anchored Tris(Aryloxide) Ligand
Palumbo CT, et al
Inorganic Chemistry, 57(5), 2823-2833 (2018)
Using Diamagnetic Yttrium and Lanthanum Complexes to Explore Ligand Reduction and C?H Bond Activation in a Tris(aryloxide)mesitylene Ligand System
Palumbo CT, et al.
Inorganic Chemistry, 57(20), 12876?12884-12876?12884 (2018)
Nitro Compounds, Aromatic
Ullmann's Encyclopedia of Industrial Chemistry (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service