Skip to Content
Merck
All Photos(1)

Key Documents

86477

Supelco

(+)-Terpinen-4-ol

analytical standard

Synonym(s):

(S)-1-Isopropyl-4-methyl-3-cyclohexen-1-ol, (S)-p-Menth-1-en-4-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein:
2411938
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (sum of enantiomers, GC)

form

(Liquid or Viscous Liquid)

optical purity

enantiomeric ratio: ~2:1

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.479

bp

211-213 °C (lit.)

density

0.933 g/mL at 20 °C (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

CC(C)[C@]1(O)CCC(C)=CC1

InChI

1S/C10H18O/c1-8(2)10(11)6-4-9(3)5-7-10/h4,8,11H,5-7H2,1-3H3/t10-/m1/s1

InChI key

WRYLYDPHFGVWKC-SNVBAGLBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(+)-Terpinen-4-ol is an important constituent of tea tree oil, possessing antimycotic activity, which can be extracted from the leaves and flowers of Melaleuca alternifolia.

Application

(+)-Terpinen-4-ol has been used as a standard in studying the effect of tea tree oil extracted from Melaleuca. Alternifolia tree on cereal pathogenic fungi like Fusarium graminearum, Fusarium culmorum and Pyrenophora graminea.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Highly purified monoterpene. Chiral starting material for many hydroxylation and epoxidation reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P.W.D. Mitchell
Organic preparations and procedures international, 22, 534-534 (1990)
[On the aldolase in rabbit liver: molecular weight, dissociation into sub-units. On aldolases. 5].
P Christen et al.
Helvetica chimica acta, 48(5), 1050-1056 (1965-07-10)
In vitro antifungal activity of the tea tree (Melaleuca alternifolia) essential oil and its major components against plant pathogens
Terzi.V, et al.
Letters in Applied Microbiology, 44, 613-618 (2007)
M. Pailer et al.
Monatshefte fur Chemie / Chemical Monthly, 112, 987-987 (1981)
Nurhayat Tabanca et al.
Biomolecules, 10(5) (2020-05-02)
The Mediterranean fruit fly, Ceratitis capitata (Wiedemann) (Diptera: Tephritidae) poses a major threat to fruit and vegetable production in the United States and throughout the world. New attractants and detection methods could improve control strategies for this invasive pest. In

Protocols

Fast GC analysis of sweet orange essential oil in hexane. Key components identified includes: β-Farnesene; α-Huµlene; Germacrene D; (+)-Valencene; Bicyclogermacrene; (+)-δ-Cadinene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service