Skip to Content
Merck
All Photos(2)

Key Documents

94141

Supelco

4-Chloro-α-cyanocinnamic acid

matrix substance for MALDI-MS, ≥95.0% (HPLC)

Synonym(s):

3-(4-Chlorophenyl)-2-cyano-2-propenoic acid, 3-(4-Chlorophenyl)-2-cyanoacrylic acid, p-Chloro-α-cyanocinnamic acid, p-Chlorobenzylidenecyanoacetic acid, ClCCA, NSC 2479

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6ClNO2
CAS Number:
Molecular Weight:
207.61
Beilstein:
2616773
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

grade

matrix substance for MALDI-MS

Quality Level

Assay

≥95.0% (HPLC)

form

crystals

analyte functional class(es)

drugs of abuse, ionic liquids

analyte chemical class(es)

chlorinated lipids, lipids, peptides, phospholipids, phosphopeptides

technique(s)

MALDI-MS: suitable

solubility

methanol: 100 mg/10 mL, clear, colorless to light yellow

SMILES string

OC(=O)\C(=C\c1ccc(Cl)cc1)C#N

InChI

1S/C10H6ClNO2/c11-9-3-1-7(2-4-9)5-8(6-12)10(13)14/h1-5H,(H,13,14)/b8-5+

InChI key

MXCRRKYUQNHWLJ-VMPITWQZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • suitable as MALDI-Matrix material
  • suitable for collision-induced dissociation MS/MS (CID-MS/MS)

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Florian Baum et al.
Journal of agricultural and food chemistry, 61(38), 9110-9117 (2013-09-03)
Multiphosphorylated peptides endogenously present in milk exert anticariogenic activity due to their calcium binding capacity. This study performed comprehensive analysis of multiphosphorylated peptides in raw milk using matrix-assisted laser desorption ionization time-of-flight mass spectrometry (MALDI-TOF-MS). Since phosphopeptides are often negatively
Tiffany Porta et al.
Journal of mass spectrometry : JMS, 46(2), 144-152 (2011-01-25)
Analysis of low molecular weight compounds (LMWC) in complex matrices by vacuum matrix-assisted laser desorption/ionization (MALDI) often suffers from matrix interferences, which can severely degrade limits of quantitation. It is, therefore, useful to have available a range of suitable matrices
Maurice H J Selman et al.
Proteomics, 12(9), 1337-1348 (2012-05-17)
For MALDI analysis of glycans and glycopeptides, the choice of matrix is crucial in minimizing desialylation by mass spectrometric in-source and metastable decay. Here, we evaluated the potential of 4-chloro-α-cyanocinnamic acid (Cl-CCA) for MALDI-TOF-MS analysis of labile sialylated tryptic N-glycopeptides
Maja Pučić Baković et al.
Journal of proteome research, 12(2), 821-831 (2013-01-10)
Age and sex dependence of subclass specific immunoglobulin G (IgG) Fc N-glycosylation was evaluated for 1709 individuals from two isolated human populations. IgGs were obtained from plasma by affinity purification using 96-well protein G monolithic plates and digested with trypsin.
John D Leszyk
Journal of biomolecular techniques : JBT, 21(2), 81-91 (2010-07-02)
MALDI-TOF continues to be an important tool for many proteomic studies. Recently, a new rationally designed matrix 4-chloro-alpha-cyanocinnamic acid was introduced, which is reported to have superior performance as compared with the "gold standard" alpha-cyano-4-hydroxycinnamic acid (CHCA). In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service