Skip to Content
Merck
All Photos(1)

Key Documents

46745

Supelco

Spiramycin from Streptomyces sp.

VETRANAL®, analytical standard, mixture of isomers

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
E Number:
E710
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

quality

mixture of isomers

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

antibiotic activity spectrum

Gram-positive bacteria

application(s)

cleaning products
clinical
cosmetics
food and beverages
forensics and toxicology
personal care
pharmaceutical (small molecule)

format

neat

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

CO[C@H]1[C@H](O)CC(=O)O[C@H](C)C\C=C\C=C\[C@H](OC2CCC(C(C)O2)N(C)C)[C@H](C)C[C@H](CC=O)[C@@H]1OC3OC(C)C(OC4CC(C)(O)C(O)C(C)O4)C(C3O)N(C)C

InChI

1S/C43H74N2O14/c1-24-21-29(19-20-46)39(59-42-37(49)36(45(9)10)38(27(4)56-42)58-35-23-43(6,51)41(50)28(5)55-35)40(52-11)31(47)22-33(48)53-25(2)15-13-12-14-16-32(24)57-34-18-17-30(44(7)8)26(3)54-34/h12-14,16,20,24-32,34-42,47,49-51H,15,17-19,21-23H2,1-11H3/b13-12+,16-14+/t24-,25-,26?,27?,28?,29+,30?,31-,32+,34?,35?,36?,37?,38?,39+,40+,41?,42?,43?/m1/s1

InChI key

ACTOXUHEUCPTEW-JMRHEKERSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: macrolide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alain Berrébi et al.
American journal of obstetrics and gynecology, 203(6), 552-552 (2010-07-17)
Maternal toxoplasmosis infection acquired during pregnancy carries significant risk of fetal damage. We aimed to assess the long-term outcome of children and young adults with congenital toxoplasmosis diagnosed and treated in utero. This was a 20 year prospective study (1985-2005).
Ming-juan Wang et al.
Journal of chromatography. A, 1217(42), 6531-6544 (2010-09-17)
Investigation of acetylspiramycin (ASPM) and its related substances was carried out using a reversed-phase liquid chromatography/tandem mass spectrometry method. The identification of impurities in the ASPM complex was performed with a quadrupole ion trap mass spectrometer, with an electrospray ionization
Daniel Ajzenberg
Expert review of anti-infective therapy, 9(2), 169-171 (2011-02-24)
Every 3 years, the International Congress on Congenital Toxoplasmosis meeting gathers experts with different backgrounds who are involved in congenital toxoplasmosis: gynecologists, pediatricians, ophthalmologists, microbiologists, epidemiologists and research scientists. Most attendees come from the Americas and Europe, where substantial work
Ying Liu et al.
Ecotoxicology and environmental safety, 86, 23-30 (2012-09-29)
Cyanobacteria may interact with antibiotic contaminants in aquatic environments, but the interaction effects and mechanisms remain unclear. In the present study, aqueous culture of Microcystis aeruginosa was exposed to 50ng/l-1μg/l of spiramycin and amoxicillin for seven days. The influences of
Wai Kit Chew et al.
Antimicrobial agents and chemotherapy, 56(4), 1762-1768 (2012-01-25)
Toxoplasma gondii is a parasite that generates latent cysts in the brain; reactivation of these cysts may lead to fatal toxoplasmic encephalitis, for which treatment remains unsuccessful. We assessed spiramycin pharmacokinetics coadministered with metronidazole, the eradication of brain cysts and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service