Skip to Content
Merck
All Photos(1)

Key Documents

33586

Supelco

Flunixin

VETRANAL®, analytical standard

Synonym(s):

2-[2-Methyl-3-(trifluoromethyl)phenylamino]nicotinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11F3N2O2
CAS Number:
Molecular Weight:
296.24
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cc1c(Nc2ncccc2C(O)=O)cccc1C(F)(F)F

InChI

1S/C14H11F3N2O2/c1-8-10(14(15,16)17)5-2-6-11(8)19-12-9(13(20)21)4-3-7-18-12/h2-7H,1H3,(H,18,19)(H,20,21)

InChI key

NOOCSNJCXJYGPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Flunixin is a potent nonsteroidal anti-inflammatory drug, primarily used as an anti-inflammatory agent and analgesic for the short-term treatment of moderate pain and inflammation in farm animals. It is also effective in inhibiting the synthesis of prostaglandins.
Flunixin is classified under the group of non-steroidal anti-inflammatory drugs (NSAIDs).

Application

Flunixin may be used as a reference standard for the determination of NSAID drug, flunixin in biological samples using high-performance liquid chromatography with diode array detection (HPLC-DAD)/particle beam mass spectrometry (PB-MS)/atmospheric pressure ionization mass spectrometry (API-MS) and gas chromatography coupled to mass spectrometry (GC-MS).
Flunixin may be used as an analytical reference standard for the determination of flunixin in:
  • Bovine muscle samples by hydrophilic interaction liquid chromatography-electrospray-tandem mass spectrometry (HILIC-ESI-MS/MS) equipped with selected reaction monitoring (SRM) detection.
  • Animal tissues by dispersive-solid phase extraction (d-SPE) and enhanced matrix removal for lipids (EMR-L), followed by analysis using ultra-high performance liquid chromatography-triple quadrupole or quadrupole-time-of-flight (UHPLC-QqQ or UHPLC-Q/TOF) methods in conjunction with ESI-MS/MS operating on multiple reaction monitoring (MRM) mode of detection as well as ESI-LC-MS/MS with SRM detection mode.
  • Porcine muscle samples by ESI-LC-MS/MS with MRM detection mode.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emma Nixon et al.
Frontiers in veterinary science, 7, 82-82 (2020-03-11)
Piglet castration and tail-docking are routinely performed in the United States without analgesia. Pain medications, predominately non-steroidal anti-inflammatory drugs, are used in the EU/Canada to decrease pain associated with processing and improve piglet welfare, however, past studies have shown the
Online eluent-switching technique coupled anion-exchange liquid chromatography-ion trap tandem mass spectrometry for analysis of non-steroidal anti-inflammatory drugs in pig serum.
Chang KC, et al.
Journal of Chromatography A, 1422, 222-229 (2015)
Comparison of veterinary drug residue results in animal tissues by ultrahigh-performance liquid chromatography coupled to triple quadrupole or quadrupole-time-of-flight tandem mass spectrometry after different sample preparation methods, including use of a commercial lipid removal product.
Anumol T, et al.
Analytical and Bioanalytical Chemistry, 409(10), 2639-2653 (2017)
J E Tomlinson et al.
Equine veterinary journal, 37(1), 75-80 (2005-01-18)
Recent studies have shown that flunixin prevented recovery of equine jejunum post ischaemia. However, the use of a purported cyclooxygenase (COX)-2 preferential inhibitor, etodolac, also prevented recovery. These findings may have implications for the use of nonsteroidal anti-inflammatory drugs in
Saunders Handbook of Veterinary Drugs - E-Book (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service