Skip to Content
Merck
All Photos(1)

Key Documents

45789

Supelco

Indan

analytical standard

Synonym(s):

Hydrindene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10
CAS Number:
Molecular Weight:
118.18
Beilstein:
1904376
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.537 (lit.)

bp

176 °C (lit.)

mp

−51 °C (lit.)

density

0.965 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

C1Cc2ccccc2C1

InChI

1S/C9H10/c1-2-5-9-7-3-6-8(9)4-1/h1-2,4-5H,3,6-7H2

InChI key

PQNFLJBBNBOBRQ-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher S Frampton et al.
Acta crystallographica. Section C, Crystal structure communications, 68(Pt 8), o323-o326 (2012-08-02)
The title molecular salt, C(8)H(12)N(+)·C(26)H(21)O(3)(-), contains a dimeric indane pharmacophore that demonstrates potent anti-inflammatory activity. The indane group of the anion exhibits some disorder about the α-C atom, which appears common to many structures containing this group. A model to
Yuka Kobayashi et al.
Chirality, 17(2), 108-112 (2005-01-22)
Both novel enantiopure trans-1-aminobenz[f]indan-2-ols (4) were obtained from the racemate by the diastereomeric salt formation with (+)- and (-)-dibenzoyltartaric acids (8), respectively, and the absolute configuration of the enantiomer 4 in the less-soluble diastereomeric salt of racemic 4 with (+)-8
Konstantin Ulanenko et al.
The Journal of organic chemistry, 71(18), 7053-7056 (2006-08-26)
5-Dimethylamino-1-aminoindan undergoes thermal decomposition and reacts with 6-chlorouracil to give 5-indanyl-6-chlorouracil derivative 9. The formation of 9 may be rationalized by a putative mechanism based on the intermediacy of the imminium methide species 8a.
Stuart N L Bennett et al.
Bioorganic & medicinal chemistry letters, 20(12), 3511-3514 (2010-05-25)
A series of carboxylic acid glycogen phosphorylase inhibitors, which have potential as oral antidiabetic agents, is described. Defining and applying simple physicochemical design criteria was used to assess the opportunity and to focus synthetic efforts on compounds with the greatest
Kenji Kabashima et al.
FEBS letters, 578(1-2), 36-40 (2004-12-08)
Nuclear factor kappa B (NF-kappaB) plays a wide variety of pathophysiological roles and modulation of its pathway can be a good novel drug target. Here, we found that our recently synthesized NF-kappaB inhibitor attenuated an ovalbumin-specific delayed-type hypersensitivity response in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service