Skip to Content
Merck
All Photos(1)

Key Documents

CRM47933

Supelco

EPA 502/524 Volatiles Organic Calibration Mix A (without gases)

certified reference material, 200 μg/mL each component in methanol, ampule of 1 mL

Synonym(s):

EPA 502/524 Volatiles Organic Calibration Mix (without gases)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Agency

EPA 502
EPA 524

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

200 μg/mL each component in methanol

technique(s)

gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Benzene
    Bromobenzene
    Bromochloromethane
    Bromodichloromethane
    Bromoform
    Butylbenzene
    sec-Butylbenzene
    tert-Butylbenzene
    Carbon tetrachloride
    Chlorobenzene
    Chloroform
    2-Chlorotoluene
    4-Chlorotoluene
    Dibromochloromethane
    1,2-Dibromo-3-chloropropane
    1,2-Dibromoethane
    Dibromomethane
    1,2-Dichlorobenzene
    1,3-Dichlorobenzene
    1,4-Dichlorobenzene
    1,1-Dichloroethane
    1,2-Dichloroethane
    1,1-Dichloroethylene
    cis-1,2-Dichloroethylene
    trans-1,2-Dichloroethylene
    Dichloromethane
    1,2-Dichloropropane
    1,3-Dichloropropane
    2,2-Dichloropropane
    1,1-Dichloro-1-propene
    cis-1,3-Dichloropropene
    trans-1,3-Dichloropropene
    Ethylbenzene
    Hexachloro-1,3-butadiene
    Cumene
    p-Isopropyltoluene
    Naphthalene
    Propylbenzene
    Styrene
    1,1,1,2-Tetrachloroethane
    1,1,2,2-Tetrachloroethane
    Tetrachloroethylene
    Toluene
    1,2,3-Trichlorobenzene
    1,2,4-Trichlorobenzene
    1,1,1-Trichloroethane
    1,1,2-Trichloroethane
    Trichloroethylene
    1,2,3-Trichloropropane
    1,2,4-Trimethylbenzene
    1,3,5-Trimethylbenzene
    m-Xylene
    o-Xylene
    p-Xylene
    See All (54)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

WGK

WGK 2

Flash Point(F)

closed cup

Flash Point(C)

closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service