Merck
Novel 5-dimethylamino-1- and 2-indanyl uracil derivatives.
The Journal of organic chemistry (2006-08-26)
Konstantin Ulanenko, Eliezer Falb, Hugo E Gottlieb, Yaacov Herzig
ABSTRACT
5-Dimethylamino-1-aminoindan undergoes thermal decomposition and reacts with 6-chlorouracil to give 5-indanyl-6-chlorouracil derivative 9. The formation of 9 may be rationalized by a putative mechanism based on the intermediacy of the imminium methide species 8a.
Sigma-Aldrich
Indan, 95%
Supelco
Indan, analytical standard