Skip to Content
Merck
All Photos(1)

Key Documents

PHR1537

Supelco

Oxytetracycline

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Oxytetracycline dihydrate, OTC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O9 · 2H2O
CAS Number:
Molecular Weight:
496.46
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 782
traceable to Ph. Eur. Y0001896
traceable to USP 1491004

API family

oxytetracycline

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

-10 to -25°C

SMILES string

O.[H][C@@]12[C@@H](O)[C@@]3([H])C(C(=O)c4c(O)cccc4[C@@]3(C)O)=C(O)[C@]1(O)C(=O)C(C(N)=O)=C(O)[C@H]2N(C)C

InChI

1S/C22H24N2O9.H2O/c1-21(32)7-5-4-6-8(25)9(7)15(26)10-12(21)17(28)13-14(24(2)3)16(27)11(20(23)31)19(30)22(13,33)18(10)29;/h4-6,12-14,17,25,27-29,32-33H,1-3H3,(H2,23,31);1H2/t12-,13-,14+,17+,21-,22+;/m1./s1

InChI key

IBZHEBHGZFICKS-IFLJXUKPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Oxytetracycline is a broad-spectrum antibiotic which finds a wide use in veterinary medicine because of its potential to inhibit the protein synthesis in gram-positive and gram-negative bacteria. It can be used on animal species like cattle, sheep, goats and pigs.

Application

Oxytetracycline may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography and spectrofluorimetry techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Antibiotic produced by Streptomyces rimosus.
Mode of Action: Inhibits protein synthesis (elongation) by preventing binding of aminoacyl-tRNA to the 30S subunit.
Antimicrobial spectrum: Gram-negative and Gram-positive bacteria.
Mode of Resistance: Active efflux, ribosome protection, tetracycline inactivation.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC0364 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative analysis for oxytetracycline in medicated premixes and feeds by second-derivative synchronous spectrofluorimetry.
Fernandez-Gonzalez R, et al.
Analytica Chimica Acta, 455(1), 143-148 (2002)
Spectrophotometric determination of hydrocortisone, nystatin and oxytetracycline in synthetic and pharmaceutical preparations based on various univariate and multivariate methods.
Gallego JML and Arroyo JP
Analytica Chimica Acta, 460(1), 85-97 (2002)
C18 columns for the simultaneous determination of oxytetracycline and its related substances by reversed-phase high performance liquid chromatography and UV detection.
Smyrniotakis CG and Archontaki HA
Journal of Pharmaceutical and Biomedical Analysis, 43(2), 506-514 (2007)
Chris Christou et al.
PloS one, 9(12), e114122-e114122 (2014-12-03)
The healing of critical sized segmental defects is an ongoing clinical problem. No method has achieved pre-eminence. The Masquelet technique is a relatively new innovation involving the induction of a fibrous tissue membrane around the bone defect site taking advantage
Achilleas G Mitrakas et al.
Anti-cancer drugs, 25(10), 1182-1191 (2014-07-19)
Abnormal mitochondrial function is common in cancer cells and activates metabolic pathways suppressed in normal tissues. Experimental and clinical studies suggest that mitochondria might serve as targets for novel anticancer therapies. We investigated whether mitochondrial metabolism-interfering agents (MMIAs) available currently

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service