Skip to Content
Merck
All Photos(1)

Key Documents

32054

Supelco

Captan

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8Cl3NO2S
CAS Number:
Molecular Weight:
300.59
Beilstein:
234872
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

ClC(Cl)(Cl)SN1C(=O)C2CC=CCC2C1=O

InChI

1S/C9H8Cl3NO2S/c10-9(11,12)16-13-7(14)5-3-1-2-4-6(5)8(13)15/h1-2,5-6H,3-4H2

InChI key

LDVVMCZRFWMZSG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Captan, an agricultural fungicide, reacts with sulfhydryl group to release thiophosgenes which can denature proteins. It has carcinogenic and teratogenic properties which can cause oral toxicity to birds and mammals.

Application

Captan has been used as reference standard in HPLC analysis to determine captan concentration from tablets.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Acute 1 - Carc. 2 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Transgenic strains of the nematode C. elegans in biomonitoring and toxicology: effects of captan and related compounds on the stress response.
Jones D
Toxicology, 109(2-3), 119-127 (1996)
High-performance liquid chromatographic determination of sulphur and captan in a mixture.
Fedeli G
Journal of Chromatography A, 447(1), 263-267 (1988)
Punam Pahwa et al.
Journal of agromedicine, 17(1), 40-50 (2011-12-24)
The objective of this study was to investigate the putative associations of specific pesticides with multiple myeloma. A matched, population-based, case-control study was conducted among men residing in six Canadian provinces (Quebec, Ontario, Manitoba, Saskatchewan, Alberta, and British Columbia). Data
Roberto Heredia-Ortiz et al.
Toxicology letters, 213(1), 27-34 (2011-10-08)
Measurement of tetrahydrophthalimide (THPI) in urine has been used for the biomonitoring of exposure to the widely used captan fungicide in workers. To allow a better understanding of the toxicokinetics of captan and its key biomarker of exposure, a human
Anil Kumar Singh et al.
Applied biochemistry and biotechnology, 164(1), 77-88 (2010-11-05)
A 56.56-kDa extracellular chitinase from Paenibacillus sp. D1 was purified to 52.3-fold by ion exchange chromatography using SP Sepharose. Maximum enzyme activity was recorded at pH 5.0 and 50 °C. MALDI-LC-MS/MS analysis identified the purified enzyme as chitinase with 60% similarity to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service