Skip to Content
Merck
All Photos(1)

Key Documents

45681

Supelco

Tetramethrin

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H25NO4
CAS Number:
Molecular Weight:
331.41
Beilstein:
8807938
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

C\C(C)=C\C1C(C(=O)OCN2C(=O)C3=C(CCCC3)C2=O)C1(C)C

InChI

1S/C19H25NO4/c1-11(2)9-14-15(19(14,3)4)18(23)24-10-20-16(21)12-7-5-6-8-13(12)17(20)22/h9,14-15H,5-8,10H2,1-4H3

InChI key

CXBMCYHAMVGWJQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - STOT SE 2 Inhalation

Target Organs

Nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J H Song et al.
Brain research, 712(2), 258-264 (1996-03-18)
The differential effects of the pyrethroid tetramethrin on tetrodotoxin-sensitive (TTX-S) and tetrodotoxin-resistant (TTX-R) single sodium channel currents in rat dorsal root ganglion (DRG) neurons were investigated using the outside-out configuration of patch-clamp technique. Channel conductances were 10.7 and 6.3 pS
H Motomura et al.
Neurotoxicology, 22(3), 329-339 (2001-07-18)
Type I and type II pyrethroids are known to modulate the sodium channel to cause persistent openings during depolarization and upon repolarization. Although there are some similarities between the two types of pyrethroids in their actions on sodium channels, the
I V Tabarean et al.
The Journal of pharmacology and experimental therapeutics, 299(3), 988-997 (2001-11-21)
Pyrethroid insecticides may be classified into two groups: type I pyrethroids lack a cyano group in the alpha-position, whereas type II pyrethroids have a cyano group. Both types prolong the sodium channel current thereby causing hyperexcitability, yet details of modulation
Ali Niazi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 69(4), 1165-1169 (2007-07-27)
The simultaneous determination of cypermethrin and tetramethrin mixtures by using spectrophotometric method is a difficult problem in analytical chemistry, due to spectral interferences. By multivariate calibration methods, such as partial least squares (PLS) regression, it is possible to obtain a
Y Tomigahara et al.
Xenobiotica; the fate of foreign compounds in biological systems, 24(12), 1205-1214 (1994-12-01)
1. To examine the metabolic fate of (1RS, trans)- or (1RS, cis)-tetramethrin [3, 4, 5, 6-tetrahydrophthalimidomethyl (1RS, trans)- or (1RS, cis)-chrysanthemate], rat was administered a single oral dose of trans- or cis-[alcohol-14C]tetramethrin at dose levels of 2 or 250 mg/kg.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service