Skip to Content
Merck
All Photos(2)

Key Documents

33309

Supelco

Esbiothrin

PESTANAL®, analytical standard

Synonym(s):

3-Allyl-2-methyl-4-oxo-2-cyclopentenyl chrysanthemate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26O3
CAS Number:
Molecular Weight:
302.41
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

SMILES string

C\C(C)=C\[C@@H]1[C@@H](C(=O)O[C@H]2CC(=O)C(CC=C)=C2C)C1(C)C

InChI

1S/C19H26O3/c1-7-8-13-12(4)16(10-15(13)20)22-18(21)17-14(9-11(2)3)19(17,5)6/h7,9,14,16-17H,1,8,10H2,2-6H3

InChI key

ZCVAOQKBXKSDMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Esbiothrin is a synthetic pyrethroid, which is widely used against household pests and in public health.

Application

Esbiothrin may be used as a reference standard for the determination of the analyte in electric-mosquito coils using gas chromatography (GC) technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sublethal toxicity of esbiothrin relationship with total antioxidant status and in vivo genotoxicity assessment in fish (Cyprinus carpio L., 1758) using the micronucleus test and comet assay.
Selvi M, et al.
Environmental Toxicology, 28(11), 644-651 (2011)
Golla Madhubabu et al.
Inhalation toxicology, 24(3), 143-152 (2012-02-24)
Continuous inhalation of allethrin-based mosquito coil smoke may affect fertility, an aspect that has not received much attention. In this study, we attempt to understand the harmful effects on the male reproductive system caused by continuous exposure to allethrin-based mosquito
Koen Desmet et al.
Chemosphere, 71(11), 2193-2198 (2008-04-02)
Spatial concentration distribution of a chemical in an indoor environment is an important factor in the evaluation of chemical nuisances. However, straightforward techniques for the determination of this distribution are not very common and usually limited in their application. Sorptive
April P Neal et al.
Toxicological sciences : an official journal of the Society of Toxicology, 116(2), 604-613 (2010-05-15)
Pyrethroid insecticides are one of the most widely used classes of insecticides. Previous studies revealed that pyrethroids potently affect the insect voltage-gated sodium (Na(+)) channel (VGSC), resulting in prolonged channel open time. However, recent findings have suggested that pyrethroids may
Mahmut Selvi et al.
Environmental toxicology, 28(11), 644-651 (2011-09-14)
Esbiothrin, synthetic pyrethroid with quick activity against insects, is widely used against household pests and in public health. Despite widespread use, data on ecotoxicity and genotoxic effects are extremely scarce. The aim of the present study is to evaluate the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service