Skip to Content
Merck
All Photos(1)

Key Documents

45680

Supelco

Tetradifon

PESTANAL®, analytical standard

Synonym(s):

2,4,4′,5-Tetrachlorodiphenyl sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H6Cl4O2S
CAS Number:
Molecular Weight:
356.05
Beilstein:
2292528
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Clc1ccc(cc1)S(=O)(=O)c2cc(Cl)c(Cl)cc2Cl

InChI

1S/C12H6Cl4O2S/c13-7-1-3-8(4-2-7)19(17,18)12-6-10(15)9(14)5-11(12)16/h1-6H

InChI key

MLGCXEBRWGEOQX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetradifon is a fungicide, commonly used for control of red spider mites affecting citrus fruits, deciduous fruits and ornamental plants. It is also used for weed control management.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tetradifon may be used as an analytical reference standard for the quantification of the analyte in fruit juice samples and essential oils of citrus fruits using different chromatography techniques.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mites Injurious to Economic Plants (1975)
Analysis of pesticide residues in essential oils of citrus fruit by GC-MS and HPLC-MS after solid-phase extraction
Barrek S, et al.
Journal of Separation Science, 376(2), 157-161 (2003)
Simultaneous determination of various pesticides in fruit juices by HPLC-DAD
Topuz S, et al.
Food Control, 16(1), 87-92 (2005)
M J Villarroel et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 35(2), 211-227 (2000-03-29)
Two Daphnia magna offsprings (animals from the first and third brood) whose parentals (F0-generation) were exposed during 21 days to different tetradifon (4-chlorophenyl 2,4,5-trichlophenyl sulfone) concentrations, were transferred to a pesticide free medium during 21 days (recovery period). The algae
M J Villarroel et al.
Ecotoxicology and environmental safety, 44(1), 40-46 (1999-09-29)
The feeding behavior of the cladocera Daphnia magna subjected to a short-term exposure to the acaricide tetradifon (4-chlorophenyl 2,4, 5-trichlorophenyl sulfone) was studied. The experiments were performed using the unicellular algae Nannochloris oculata at a density of 5x10(5) cells/ml as

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service