Skip to Content
Merck
All Photos(1)

Key Documents

45642

Supelco

Propiconazole

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17Cl2N3O2
CAS Number:
Molecular Weight:
342.22
Beilstein:
9349305
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

description

mixture of stereo isomers

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCCC1COC(Cn2cncn2)(O1)c3ccc(Cl)cc3Cl

InChI

1S/C15H17Cl2N3O2/c1-2-3-12-7-21-15(22-12,8-20-10-18-9-19-20)13-5-4-11(16)6-14(13)17/h4-6,9-10,12H,2-3,7-8H2,1H3

InChI key

STJLVHWMYQXCPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propiconazole is a systemic fungicide and its mode of action involves the inhibition of ergosterol biosynthesis.

Application

Propiconazole may be used as an analytical reference standard for the quantification of the analyte in must, wine samples and environmental samples using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

392.0 °F - Pensky-Martens closed cup

Flash Point(C)

200 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

70th International Symposium on Molecular Spectroscopy (2004)
HPLC method for simultaneous determination of fungicides: carbendazim, metalaxyl, folpet, and propiconazole in must and wine
Lopez F.L, et al.
Journal of Agricultural and Food Chemistry, 37(3), 684-687 (1989)
Jet C Van De Steene et al.
Journal of the American Society for Mass Spectrometry, 19(5), 713-718 (2008-03-18)
When developing an LC-MS/MS-method matrix effects are a major issue. The effect of co-eluting compounds arising from the matrix can result in signal enhancement or suppression. During method development much attention should be paid to diminishing matrix effects as much
Barbara S Shane et al.
Environmental and molecular mutagenesis, 53(1), 1-9 (2012-02-14)
Propiconazole (PPZ) is a conazole fungicide that is not mutagenic, clastogenic, or DNA damaging in standard in vitro and in vivo genetic toxicity tests for gene mutations, chromosome aberrations, DNA damage, and cell transformation. However, it was demonstrated to be
Maryline Calonne et al.
Chemosphere, 87(4), 376-383 (2012-01-14)
The increasing concentrations impact (0.02, 0.2 and 2 mg L(-1)) of a Sterol Biosynthesis Inhibitor (SBI) fungicide, propiconazole, was evaluated on development and sterol metabolism of two non-target organisms: mycorrhizal or non-mycorrhizal transformed chicory roots and the arbuscular mycorrhizal fungus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service