Skip to Content
Merck
All Photos(2)

Key Documents

42038

Supelco

2,3-Butanediol

analytical standard, mixture of racemic and meso forms

Synonym(s):

2,3-Butylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH(OH)CH3
CAS Number:
Molecular Weight:
90.12
Beilstein:
969165
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.433 (lit.)
n20/D 1.438

bp

183-184 °C (lit.)

mp

25 °C (lit.)

density

1.002 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(O)C(C)O

InChI

1S/C4H10O2/c1-3(5)4(2)6/h3-6H,1-2H3

InChI key

OWBTYPJTUOEWEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3-Butanediol is a chiral compound which is extensively utilized in chemical feedstocks and liquid fuels.

Application

2,3-Butanediol may be used as a reference standard for the determination of 2,3-butanediol in:
  • ′Fino′ sherry wine by rotatory and continuous liquid−liquid extraction (LLE) and headspace solid-phase microextraction (HS-SPME) combined with gas chromatography-mass spectrometry (GC-MS).
  • Cell cultures as well as fermentation broths by high performance anion-exchange chromatography with pulsed amperometric detection (HPAE-PAD).
  • Cocoa (Theobroma cacao L.) by HS-SPME-GC-MS with principal component analysis (PCA).
  • Vinegar samples by HS-SPME combined with GC-MS in electron impact ionization (EI+) mode.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dynamics of volatile and non-volatile compounds in cocoa (Theobroma cacao L.) during fermentation and drying processes using principal components analysis
Rodriguez-Campos J, et al.
Food Research International, 44(1), 250-258 (2011)
Determination of carbohydrates, sugar alcohols, and glycols in cell cultures and fermentation broths using high-performance anion-exchange chromatography with pulsed amperometric detection
Hanko VP and Rohrer JS
Analytical Biochemistry, 283(2), 192-199 (2000)
Headspace solid-phase microextraction analysis of aroma compounds in vinegar: validation study
Marin RN, et al.
Journal of Chromatography A, 967(2), 261-267 (2002)
Comparative analysis of volatile compounds of `fino?sherry wine by rotatory and continuous liquid-liquid extraction and solid-phase microextraction in conjunction with gas chromatography-mass spectrometry
Castro R, et al.
Analytica Chimica Acta, 513(1), 141-150 (2004)
Bioconversion of pentoses to 2,3-butanediol by Klebsiella pneumoniae.
N B Jansen et al.
Advances in biochemical engineering/biotechnology, 27, 85-99 (1983-01-01)

Chromatograms

suitable for GC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service