Skip to Content
Merck
All Photos(1)

Key Documents

40127-U

Supelco

Acetoin

analytical standard

Synonym(s):

3-Hydroxy-2-butanone, Acetylmethylcarbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH(OH)CH3
CAS Number:
Molecular Weight:
88.11
Beilstein:
385636
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

CofA

current certificate can be downloaded

quality

monomer

packaging

pkg of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.417 (lit.)

bp

148 °C (lit.)

mp

15 °C (monomer)
90 °C (dimer) (lit.)

density

1.013 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CC(O)C(C)=O

InChI

1S/C4H8O2/c1-3(5)4(2)6/h3,5H,1-2H3

InChI key

ROWKJAVDOGWPAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcello R Silveira et al.
Food chemistry, 279, 120-127 (2019-01-07)
The effect of cold plasma processing time and gas flow on bioactive compounds such as vitamin C, carotenoids and phenolic compounds, DPPH, angiotensin-converting-enzyme (ACE) inhibitory activity, fatty acids profile, and volatile compounds of guava-flavored whey beverage was investigated. For comparative
Zhen Liu et al.
Bioresource technology, 102(22), 10741-10744 (2011-09-29)
Production of highly pure (2S,3S)-2,3-butanediol ((2S,3S)-2,3-BD) and (3S)-acetoin ((3S)-AC) in high concentrations is desirable but difficult to achieve. In the present study, glucose was first transformed to a mixture of (2S,3S)-2,3-BD and meso-2,3-BD by resting cells of Klebsiella pneumoniae CICC
Ranjita Biswas et al.
Applied microbiology and biotechnology, 94(3), 651-658 (2012-03-01)
Production of 2,3-butanediol by Bacillus subtilis takes place in late-log or stationary phase, depending on the expression of bdhA gene encoding acetoin reductase, which converts acetoin to 2,3-butanediol. The present work focuses on the development of a strain of B.
Maria del Pilar Marquez-Villavicencio et al.
PloS one, 6(8), e22974-e22974 (2011-08-31)
Pectobacterium species are necrotrophic bacterial pathogens that cause soft rot diseases in potatoes and several other crops worldwide. Gene expression data identified Pectobacterium carotovorum subsp. carotovorum budB, which encodes the α-acetolactate synthase enzyme in the 2,3-butanediol pathway, as more highly
M Otsuka et al.
Journal of chromatography, 577(2), 215-220 (1992-06-10)
Acetoin was quantitatively oxidized into diacetyl by Fe3+ in 1 M perchloric acid. The reaction of diacetyl with 4,5-dichloro-1,2-diaminobenzene afforded 6,7-dichloro-2,3-dimethylquinoxaline (DCDMQ), which was extracted by benzene containing aldrin (25 ng/ml) as an internal standard, and determined by gas chromatography

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service