Skip to Content
Merck
All Photos(4)

Key Documents

A17951

Sigma-Aldrich

Acetoin

May exist as crystalline dimer

Synonym(s):

3-Hydroxy-2-butanone, Acetylmethylcarbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH(OH)CH3
CAS Number:
Molecular Weight:
88.11
Beilstein:
385636
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

form

crystals

refractive index

n20/D 1.417 (lit.)

bp

148 °C (lit.)

mp

15 °C (monomer)
90 °C (dimer) (lit.)

density

1.013 g/mL at 25 °C (lit.)

food allergen

no known allergens

storage temp.

2-8°C

SMILES string

CC(O)C(C)=O

InChI

1S/C4H8O2/c1-3(5)4(2)6/h3,5H,1-2H3

InChI key

ROWKJAVDOGWPAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ranjita Biswas et al.
Applied microbiology and biotechnology, 94(3), 651-658 (2012-03-01)
Production of 2,3-butanediol by Bacillus subtilis takes place in late-log or stationary phase, depending on the expression of bdhA gene encoding acetoin reductase, which converts acetoin to 2,3-butanediol. The present work focuses on the development of a strain of B.
M Otsuka et al.
Journal of chromatography, 577(2), 215-220 (1992-06-10)
Acetoin was quantitatively oxidized into diacetyl by Fe3+ in 1 M perchloric acid. The reaction of diacetyl with 4,5-dichloro-1,2-diaminobenzene afforded 6,7-dichloro-2,3-dimethylquinoxaline (DCDMQ), which was extracted by benzene containing aldrin (25 ng/ml) as an internal standard, and determined by gas chromatography
Maria del Pilar Marquez-Villavicencio et al.
PloS one, 6(8), e22974-e22974 (2011-08-31)
Pectobacterium species are necrotrophic bacterial pathogens that cause soft rot diseases in potatoes and several other crops worldwide. Gene expression data identified Pectobacterium carotovorum subsp. carotovorum budB, which encodes the α-acetolactate synthase enzyme in the 2,3-butanediol pathway, as more highly
Zhen Liu et al.
Bioresource technology, 102(22), 10741-10744 (2011-09-29)
Production of highly pure (2S,3S)-2,3-butanediol ((2S,3S)-2,3-BD) and (3S)-acetoin ((3S)-AC) in high concentrations is desirable but difficult to achieve. In the present study, glucose was first transformed to a mixture of (2S,3S)-2,3-BD and meso-2,3-BD by resting cells of Klebsiella pneumoniae CICC
Pieter Moons et al.
Applied and environmental microbiology, 77(10), 3422-3427 (2011-03-29)
During fermentation of sugars, a number of bacterial species are able to switch from mixed acid production to acetoin and 2,3-butanediol production in order to avoid lethal acidification of their environment, although the regulation of this switch is only poorly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service