Skip to Content
Merck
All Photos(2)

Key Documents

40462

Supelco

Red 2G

analytical standard

Synonym(s):

Acid Red 1, Amido Naphthol Red G, Azophloxine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H13N3Na2O8S2
CAS Number:
Molecular Weight:
509.42
Beilstein:
4121767
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
E Number:
E128
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[Na+].[Na+].CC(=O)Nc1cc(cc2cc(c(\N=N\c3ccccc3)c(O)c12)S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C18H15N3O8S2.2Na/c1-10(22)19-14-9-13(30(24,25)26)7-11-8-15(31(27,28)29)17(18(23)16(11)14)21-20-12-5-3-2-4-6-12;;/h2-9,23H,1H3,(H,19,22)(H,24,25,26)(H,27,28,29);;/q;2*+1/p-2/b21-20+;;

InChI key

WXLFIFHRGFOVCD-SERMZQFOSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Red 2G (E-128) is used for the quality control in food products. It belongs to the class of water-soluble synthetically prepared, food color additives, reportedly banned by the European and United states union. These synthetic food colors are commonly used to compensate for the loss of natural colors of food, destroyed during processing and storage. They also maintain and improve the color appearance in food products. E-128 is a mono azo dye, reported to cause increased iron deposition in the human kidney. It finds applications as a color additive in burger meat and sausages.
Red 2G is a compound, which belongs to the azo family of dyes, generally used in textile industry.It can mainly find applications in non-linear optical studies.

Application

Red 2G may be used as an analytical reference standard for the quantification of the analyte in:
  • Drinks and candies using reversed-phase high-performance liquid chromatography with photodiode array detection (RP-HPLC-PDA).
  • Food products, beverages and water-soluble foods using reversed-phase high-performance liquid chromatography with diode array detection (RP-HPLC-DAD).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An analysis of the possibility for health implications of joint actions and interactions between food additives
Groten PJ, et al.
Regulatory Toxicology and Pharmacology, 31(1), 77-91 (2000)
Determination of 18 water-soluble artificial dyes by LC?MS in selected matrices
Martin F, et al.
Food Chemistry, 197, 1249-1255 (2016)
Investigation of optical limiting properties of Azophloxine dye using nanosecond Z-scan technique
Parvin M and Ahamed B
International Journal of ChemTech Research, 6(4), 2493-2498 (2014)
Determination of 13 synthetic food colorants in water-soluble foods by reversed-phase high-performance liquid chromatography coupled with diode-array detector
Minioti SK, et al.
Analytica Chimica Acta, 583(1), 103-110 (2007)
The effect of high-energy radiation on aqueous solution of Acid Red 1 textile dye
Foldvary.M.Cs and Wojnarovits.L
Radiation Physics and Chemistry, 76(8), 1485- 1488 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service