Skip to Content
Merck
All Photos(2)

Key Documents

214515

Sigma-Aldrich

Chromotrope FB

Dye content 50 %

Synonym(s):

Acid Red 14, Disodium 4-hydroxy-3-[(4-sulfo-1-naphthalenyl)azo]-1-naphthalenesulfonate, Mordant Blue 79

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12N2Na2O7S2
CAS Number:
Molecular Weight:
502.43
Colour Index Number:
14720
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

composition

Dye content, 50%

mp

275 °C

λmax

383 nm
515 nm (2nd)

ε (extinction coefficient)

≥14500 at 513-519 nm in H2O at 0.03 g/L
≥33000 at 214-220 nm in H2O at 0.03 g/L
≥9000 at 320-326 nm in H2O at 0.03 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].Oc1c(cc(c2ccccc12)S([O-])(=O)=O)\N=N\c3ccc(c4ccccc34)S([O-])(=O)=O

InChI

1S/C20H14N2O7S2.2Na/c23-20-15-8-4-3-7-14(15)19(31(27,28)29)11-17(20)22-21-16-9-10-18(30(24,25)26)13-6-2-1-5-12(13)16;;/h1-11,23H,(H,24,25,26)(H,27,28,29);;/q;2*+1/p-2/b22-21+;;

InChI key

YSVBPNGJESBVRM-ZPZFBZIMSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Chromotrope FB has been used in indirect competitive CLIA (icCLIA) to detect its presence in foods.

Biochem/physiol Actions

Chromotrope FB, also known as acid red 14 (AR14), is a bright orange-red powder. It belongs to the class of monoazides. Chromotrope FB efficiently stains fatty substances and is also used to dye food and medicines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>437.0 °F

Flash Point(C)

> 225 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wayne Luallen
Tying the Classic Salmon Fly: A Modern Approach to Traditional Techniques (1997)
Meenakshi Tripathi et al.
Journal of AOAC International, 87(3), 657-663 (2004-08-04)
A simple method has been developed for the extraction, separation, and determination of synthetic colors in ice cream samples. The process involves the breakdown of emulsion by neutral detergents (Triton X-100 and Tween 20) followed by extraction with petroleum ether
L M Flaminio et al.
Pharmacological research communications, 20(10), 907-917 (1988-10-01)
14C-Carmoisine (250/ug; 1.25 x 10(6) dpm) was incubated under strictly anaerobic conditions with resting cell suspension from stool specimens collected from male rats and human male healthy adults. The kinetics of azoreduction was determined as amount of naphthionic acid (NA)
E Tragni et al.
Journal of applied toxicology : JAT, 5(5), 273-276 (1985-10-01)
Pregnant rats received 14C-Carmoisine (200 mg kg-1; 25 microCi) by gavage on days 16-19 of gestation. The animals were killed and maternal tissues, amniotic fluid, placentae, foetal membranes and foetuses were analyzed for radioactivity. No evidence for the transplacental transfer
Marietta R Issidorides et al.
Progress in neuro-psychopharmacology & biological psychiatry, 28(5), 829-847 (2004-09-15)
Our histochemical and ultrastructural studies have identified, in human catecholamine locus coeruleus (LC) neurons, abundant and large spherical protein bodies (PB), containing histone-like, arginine-rich proteins, which originate as dense bodies in mitochondria. This species-specific phenotype in the neurons of man

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service