Skip to Content
Merck
All Photos(1)

Key Documents

398179

Sigma-Aldrich

Diethanolamine

ACS reagent, ≥98.5%

Synonym(s):

2,2′-Iminodiethanol, Bis(2-hydroxyethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HN(CH2CH2OH)2
CAS Number:
Molecular Weight:
105.14
Beilstein:
605315
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

vapor density

3.6 (vs air)

vapor pressure

<0.98 atm ( 100 °C)

Assay

≥98.5%

form

solid or liquid

autoignition temp.

689 °F

eq. wt.

104.0-106.0 Apparent

expl. lim.

10.6 %

impurities

≤0.15% water
≤1.0% C2H7NO
≤1.0% C6H15NO3

ign. residue

≤0.005%

color

APHA: ≤15

refractive index

n20/D 1.477 (lit.)

bp

217 °C/150 mmHg (lit.)

mp

28 °C (lit.)

solubility

water: soluble 105 g/L at 20 °C

density

1.097 g/mL at 25 °C (lit.)

functional group

amine
hydroxyl

SMILES string

OCCNCCO

InChI

1S/C4H11NO2/c6-3-1-5-2-4-7/h5-7H,1-4H2

InChI key

ZBCBWPMODOFKDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethanolamine (DEA) is an amino alcohol commonly used in the preparation of soaps and surfactants, in agricultural chemicals and in textile processing. It is used as an absorbent for capturing CO2. Its toxic and carcinogenic effect has been studied.

Application

Diethanolamine may be used in the following processes:
  • Preparation of disodiumiminodiacetic acid by dehydrogenation.
  • As a solvent and reactant in triazine based dendrimer synthesis.
  • Preparation of diethanolamine ionic liquids employed in green Heck reaction.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Oral

Target Organs

Kidney,Liver,Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diethanolamine and N,N-diethylethanolamine ionic liquids as precatalyst-precursors and reaction media in green Heck reaction protocol.
Petrovic ZD, et al.
J. Mol. Catal. A: Chem., 327(1), 45-50 (2010)
Monitoring of Chemical Speciation of DEA-CO2-Water System by Raman Spectroscopy.
Shahid MZ, et al.
Advanced Materials Research, 1113, 358-363 (2015)
Development, Characterization of Hydroxyl Terminated Dendritic Macromolecules as Prospective Drug Carriers.
Gajjar D, et al.
American Journal of Polymer Science & Engineering, 3(1), 73-89 (2015)
A continuous diethanolamine dehydrogenation fixed bed catalyst and reactor system.
Hickman DA, et al.
Chemical Engineering Journal, 278, 447-453 (2015)
Siou-Yin Chen et al.
Inorganic chemistry, 51(8), 4448-4457 (2012-04-05)
The synthesis, X-ray crystallography, magnetic properties, and high-field electron paramagnetic resonance (HFEPR) of a new heptanuclear manganese complex [Mn(7)(heamp)(6)](ClO(4))(2)·4CH(2)Cl(2)·H(2)O (complex 2), in which heampH(3) is 2-[N,N-di(2-hydroxyethyl)aminomethyl]phenol (compound 1), is reported. Complex 2 has a hexagonal, disk-shaped topology and contains six

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service