Skip to Content
Merck
All Photos(1)

Key Documents

471828

Sigma-Aldrich

N-Methyldiethanolamine

≥99%

Synonym(s):

2,2′-Methyliminodiethanol, N,N-Bis(2-hydroxyethyl)methylamine, MDEA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3N(CH2CH2OH)2
CAS Number:
Molecular Weight:
119.16
Beilstein:
1734441
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4 (vs air)

Quality Level

vapor pressure

0.01 mmHg ( 20 °C)

Assay

≥99%

autoignition temp.

770 °F

expl. lim.

99 %, 55 °F
~56 %, 23 °F
~8.8 %

refractive index

n20/D 1.469 (lit.)

bp

246-248 °C (lit.)

density

1.038 g/mL at 25 °C (lit.)

SMILES string

CN(CCO)CCO

InChI

1S/C5H13NO2/c1-6(2-4-7)3-5-8/h7-8H,2-5H2,1H3

InChI key

CRVGTESFCCXCTH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Methyldiethanolamine is a tertiary amine utilized in boronate ester formation and nucleophilic substitutions.

Application

N-Methyldiethanolamine (MDEA) may be employed in the following studies:
  • Synthesis of aluminophosphate-based molecular sieves.
  • Preparation of N-methyl-N-R-N,N-bis(2-hydroxyethyl) ammonium bromides.
  • Preparation of cationic polyurethane dispersions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dehua Wang et al.
Journal of colloid and interface science, 445, 119-126 (2015-01-24)
In the present study, N-methyldiethanolamine (MDEA) is demonstrated to be a multifunctional structure-directing agent for the synthesis of aluminophosphate-based molecular sieves. Four types of molecular sieves, including SAPO-34, -35, AlPO-9 and -22, are for the first time acquired with MDEA
N-Methyldiethanolamine
Saher HS
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2001)
Wen-Shuai Liu et al.
Chemical biology & drug design, 85(1), 91-97 (2014-09-13)
Five N-methyl-N-R-N,N-bis(2-hydroxyethyl) ammonium bromides (R = -benzyl (chloride, BNQAS), -dodecyl (C12QAS), -tetradecyl (C14QAS), -hexadecyl (C16QAS), -octadecyl (C18QAS)) were prepared based on N-methyldiethanolamine (MDEA) and halohydrocarbon. Five QAS were characterized by FTIR, NMR, and MS. BNQAS, C12QAS, C14QAS, and C16QAS were
Highly stretchable nanoalginate based polyurethane elastomers.
Daemi H, et al.
Carbohydrate Polymers, 95(2), 630-636 (2013)
Jacob D McDonald et al.
Environmental science & technology, 48(18), 10821-10828 (2014-08-29)
Carbon dioxide (CO2) absorption with aqueous amine solvents is a method of carbon capture and sequestration (CCS) from flue gases. One concern is the possible release of amine solvents and degradation products into the atmosphere, warranting evaluation of potential pulmonary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service