Skip to Content
Merck
All Photos(1)

Key Documents

M0120000

Malathion

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19O6PS2
CAS Number:
Molecular Weight:
330.36
Beilstein:
1804525
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

malathion

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCOC(=O)CC(SP(=S)(OC)OC)C(=O)OCC

InChI

1S/C10H19O6PS2/c1-5-15-9(11)7-8(10(12)16-6-2)19-17(18,13-3)14-4/h8H,5-7H2,1-4H3

InChI key

JXSJBGJIGXNWCI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Malathion EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Baljinder Singh et al.
Critical reviews in microbiology, 40(2), 146-154 (2013-02-28)
Organophosphorus pesticide, malathion, is used in public health, residential, and agricultural settings worldwide to control the pest population. It is proven that exposure to malathion produce toxic effects in humans and other mammals. Due to high toxicity, studies are going
Shereene Idriss et al.
Journal of drugs in dermatology : JDD, 8(8), 715-720 (2009-08-12)
Malathion is an under-recognized and under-utilized therapy for head lice and scabies largely due to misperceptions about its safety profile. Specifically, its pure form as it exists in pharmaceutical preparations is non-toxic to humans in the low doses available. While
Droplet size characterization of three aerial malathion spray programs.
T Younglove et al.
Bulletin of environmental contamination and toxicology, 53(4), 493-500 (1994-10-01)
Distribution, transport, and fate of the insecticides malathion and parathion in the environment.
M S Mulla et al.
Residue reviews, 81, 1-172 (1981-01-01)
Biological and environmental impacts of the insecticides malathion and parathion on nontarget biota in aquatic ecosystems.
M S Mulla et al.
Residue reviews, 78, 100-135 (1981-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service