Skip to Content
Merck
All Photos(1)

Key Documents

34314

Supelco

Bifenthrin

PESTANAL®, analytical standard, mixture of isomers

Synonym(s):

3-[(1Z)-2-Chloro-3,3,3-trifluoro-1-propenyl]-2,2-dimethylcyclopropanecarboxylic acid (2-methylbiphenyl-3-yl)methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H22ClF3O2
CAS Number:
Molecular Weight:
422.87
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC1(C)C(C(OCC2=C(C)C(C3=CC=CC=C3)=CC=C2)=O)C1/C=C(Cl)/C(F)(F)F

InChI

1S/C23H22ClF3O/c1-14-16(10-7-11-17(14)15-8-5-4-6-9-15)12-19(28)21-18(22(21,2)3)13-20(24)23(25,26)27/h4-11,13,18,21H,12H2,1-3H3/b20-13+

InChI key

URDFVRKNOVHDTP-DEDYPNTBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bifenthrin (BF) is a pyrethroid insecticide widely used in urban and agricultural applications.

Application

Bifenthrin may be used as an analytical reference standard for the determination of the analyte in vegetables, ground and sea water and tea samples by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Skin Sens. 1 - STOT RE 1

Target Organs

Nervous system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The effect of bifenthrin on the dopaminergic pathway in juvenile rainbow trout (Oncorhynchus mykiss).
Crago J and Schlenk D
Aquatic Toxicology (Amsterdam, Netherlands), 162, 66-72 (2015)
Microwave-assisted steam distillation for the determination of organochlorine pesticides and pyrethroids in Chinese teas.
Ji J, et al.
Talanta, 71(3), 1068-1074 (2007)
Simple, rapid solid-phase extraction procedure for the determination of ultra-trace levels of pyrethroids in ground and sea water by liquid chromatography/electrospray ionization mass spectroscopy.
Gil?Garcia MD, et al.
Rapid Communications in Mass Spectrometry, 20(16), 2395-2403 (2006)
Dissipation behavior of bifenthrin residues in tea and its brew.
Tewary DK, et al.
Food Control, 16(3), 231-237 (2005)
Determination of pyrethroids in vegetables by HPLC using continuous on-line post-elution photoirradiation with fluorescence detection.
Lopez-Lopez T, et al.
Analytica Chimica Acta, 447(1-2), 101-111 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service