Skip to Content
Merck
All Photos(1)

Key Documents

34045

Supelco

Azaconazole

PESTANAL®, analytical standard

Synonym(s):

1-[2-(2,4-Dichlorophenyl)-1,3-dioxolan-2-ylmethyl]-1H-1,2,4-triazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11Cl2N3O2
CAS Number:
Molecular Weight:
300.14
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Clc1ccc(c(Cl)c1)C3(Cn2cncn2)OCCO3

InChI

1S/C12H11Cl2N3O2/c13-9-1-2-10(11(14)5-9)12(18-3-4-19-12)6-17-8-15-7-16-17/h1-2,5,7-8H,3-4,6H2

InChI key

AKNQMEBLVAMSNZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Azaconazole has been used as an internal standard in the determination of propiconazole, a fungistatic agent in white spruce, using gas chromatography/mass spectrometry (GC/MS).

Application

Azaconazole, is an agricultural fungicide, which shows activity against wood destroying fungi and sapstain fungi. It can also play the role of a disinfectant in mushroom cultivation and in the storage of fruits and vegetables.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roberts RT and Hutson HD
Metabolic Pathways of Agrochemicals: Herbicides and plant growth regulators; (1998)
G O Rankin et al.
Toxicology, 34(1), 1-11 (1985-01-01)
Azaconazole is an experimental agricultural fungicide which has shown promise for use in controlling powdery mildew on crops and bean rust. This study examined the nephrotoxic potential of a single intraperitoneal azaconazole injection (0.4 or 0.6 mmol/kg) or daily azaconazole
Interaction of azole derivatives with cytochrome P-450 isozymes in yeast, fungi, plants and mammalian cells.
Bossche VH, et al.
Pest Management Science, 21.4, 289-306 (1987)
Raman microscopy for the quantitation of propiconazole in white spruce
Kurti E, et al.
Wood Science and Technology, 39.8, 618-629 (2005)
Bryce E Mansfield et al.
PLoS pathogens, 6(9), e1001126-e1001126 (2010-10-14)
Despite the wealth of knowledge regarding the mechanisms of action and the mechanisms of resistance to azole antifungals, very little is known about how the azoles are imported into pathogenic fungal cells. Here the in-vitro accumulation and import of Fluconazole

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service