Skip to Content
Merck

34348

Supelco

Hexaconazol

PESTANAL®, analytical standard

Synonym(s):

(±)-2-(2,4-Dichlorophenyl)-1-(1H-1,2,4-triazol-1-yl)-2-hexanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17Cl2N3O
CAS Number:
Molecular Weight:
314.21
Beilstein:
8328399
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCCCC(O)(Cn1cncn1)c2ccc(Cl)cc2Cl

InChI

1S/C14H17Cl2N3O/c1-2-3-6-14(20,8-19-10-17-9-18-19)12-5-4-11(15)7-13(12)16/h4-5,7,9-10,20H,2-3,6,8H2,1H3

InChI key

STMIIPIFODONDC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SAB4503250HPA026307SAB4503244
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

mol wt

antigen 54 kDa

mol wt

antigen 54 kDa

mol wt

-

mol wt

antigen 72 kDa

Gene Information

human ... CAMK2A(815)

Gene Information

human ... CAMK2A(815)

Gene Information

human ... CAMK2B(816)

Gene Information

human ... CAMK2B(816)

form

buffered aqueous solution

form

buffered aqueous solution

form

buffered aqueous glycerol solution

form

buffered aqueous solution

General description

Hexaconazol is a triazole systemic fungicide effective against Ascomycetes and Basidiomycetes.[1]

Application

Hexaconazol may be used as an analytical reference standard for the determination of the analyte in honey,[2] surface water samples,[3] black tea,[4] and fruits/vegetables[5] by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Halimah Muhamad et al.
Drug testing and analysis, 4 Suppl 1, 112-117 (2012-08-08)
In oil palm plantations, the fungicide hexaconazole is used to control Ganoderma infection that threatens to destroy or compromisethe palm. The application of hexaconazole is usually through soil drenching, trunk injection, or a combination of these two methods. It is
Multiresidue determination of 19 fungicides in processed fruits and vegetables by capillary gas chromatography after gel permeation chromatography.
Sannino A, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 82(2), 1229-1238 (1999)
A Kishorekumar et al.
Colloids and surfaces. B, Biointerfaces, 60(2), 207-212 (2007-08-03)
The effects of two triazole compounds, triadimefon and hexaconazole, on the growth and carbohydrate metabolism were studied in Solenostemon rotundifolius Poir., Morton plants under pot culture. Plants were treated with triadimefon at 15mg l(-1) and hexaconazole at 10mg l(-1) separately
Gas chromatographic method for the determination of hexaconazole residues in black tea.
Karthika C and Sachin PJ
Journal of Zhejiang University. Science, 9(2), 160-164 (2008)
A Kishorekumar et al.
Colloids and surfaces. B, Biointerfaces, 62(2), 307-311 (2007-12-11)
The effect of different triazole compounds, viz., triadimefon (TDM) and hexaconazole (HEX) treatments on the antioxidant metabolism of Solenostemon rotundifolius Poir., Morton plants was investigated in the present study under pot culture. Plants were treated with TDM at 15 mg

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service