Skip to Content
Merck
Azaconazole PESTANAL®, analytical standard

Azaconazole

Azaconazole PESTANAL®, analytical standard
Synonym(s):
1-[2-(2,4-Dichlorophenyl)-1,3-dioxolan-2-ylmethyl]-1H-1,2,4-triazole
Empirical Formula (Hill Notation):
C12H11Cl2N3O2
CAS Number:
Molecular Weight:
300.14
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique