Skip to Content
Merck
All Photos(2)

Key Documents

32555

Supelco

Xylazine-d6

VETRANAL®, analytical standard

Synonym(s):

2-(2,6-Dimethyl-d6-phenylamino)-5,6-dihydro-4H-thiazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12D6H10N2S
CAS Number:
Molecular Weight:
226.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

mass shift

M+6

SMILES string

[2H]C([2H])([2H])c1cccc(c1NC2=NCCCS2)C([2H])([2H])[2H]

InChI

1S/C12H16N2S/c1-9-5-3-6-10(2)11(9)14-12-13-7-4-8-15-12/h3,5-6H,4,7-8H2,1-2H3,(H,13,14)/i1D3,2D3

InChI key

BPICBUSOMSTKRF-WFGJKAKNSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Xylazine-d6 may be used as an analytical standard for the determination of xylazine in:
  • Rat brain samples by liquid-liquid extraction (LLE) and gas chromatography-mass spectrometry (GC-MS).
  • Water samples by isotope-dilution liquid chromatography-tandem mass spectrometry (LC-MS/MS).

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Measurement of ketamine and xylazine in rat brain by liquid-liquid extraction and gas chromatography-mass spectrometry.
Bonfanti E, et al.
Journal of Pharmacological and Toxicological Methods, 77, 6-9 (2016)
Elodie Bonfanti et al.
Journal of pharmacological and toxicological methods, 77, 6-9 (2015-09-13)
In human and veterinary medicine, the injectable drugs ketamine and xylazine are mainly used in combination to induce, and then maintain general anaesthesia; they also provide pain and stress relief. Some side-effects have been reported on the auditory brainstem response
Comparative behavioral toxicology with two common larval fish models: Exploring relationships among modes of action and locomotor responses.
Steele WB, et al.
The Science of the Total Environment, 640, 1587-1600 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service