Skip to Content
Merck
All Photos(1)

Key Documents

C-162

Supelco

Carfentanil Oxalate solution

(sold as 100 ug/mL in methanol as free base), certified reference material, ampule of 0.5 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H30N2O3 · C2H2O4
CAS Number:
Molecular Weight:
484.54
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 0.5 mL

manufacturer/tradename

Cerilliant®

drug control

estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

(sold as 100 ug/mL in methanol as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O=C(OC)C(CC1)(N(C2=CC=CC=C2)C(CC)=O)CCN1CCC3=CC=CC=C3.OC(C(O)=O)=O

Looking for similar products? Visit Product Comparison Guide

General description

A certified Snap-N-Spike® solution suitable for use as starting material in calibrators and controls for fentanyl LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, prescription monitoring, or forensic analysis applications. Marketed under the trade name Wildnil as a general anesthetic for large animals, carfentanil hit the illicit market in 2016 as a new NPS drug or in combination with heroin. Carfentanil, a μ-opioid receptor agonist, is also known as 4-carbomethoxyfentanyl. It is a fentanyl analog 10,000 times more potent than morphine, making it one of the most potent synthetic opioids available.

Application

The Certified Reference Mateial (CRM) in solution can also be used as follows:

  • Quantification of pharmaceutical drugs, fentanyl, and its analogs from slurries prepared using powdered drug samples by paper spray-mass spectrometry (PS-MS)
  • Evaluation of portable nano-liquid chromatography-electron ionization-mass spectrometry (nLC-EI-MS technique for the on-site identification of fentanyl and its derivatives
  • Ultra-high-performance liquid chromatographic-tandem mass spectrometric (LC-MS/MS) analysis of 28 fentanyl analogs after their liquid-liquid extraction from whole blood samples
  • Development of an optimized method for collection and extraction of 17 fentanyl and 10 of the most common fentanyl adulterants using a solid, non-porous surface followed by their measurement by liquid chromatography-tandem mass spectrometry (LC-MS/MS)

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in an ampoule, protected in a “safety can”, to ensure safe transport and handling in the lab

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service