Skip to Content
Merck
All Photos(2)

Key Documents

31141

Supelco

Prosulfocarb

PESTANAL®, analytical standard

Synonym(s):

S-Benzyl dipropylthiocarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H21NOS
CAS Number:
Molecular Weight:
251.39
Beilstein:
4804364
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCCN(CCC)C(=O)SCc1ccccc1

InChI

1S/C14H21NOS/c1-3-10-15(11-4-2)14(16)17-12-13-8-6-5-7-9-13/h5-9H,3-4,10-12H2,1-2H3

InChI key

NQLVQOSNDJXLKG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mara Gennari et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 37(4), 297-305 (2002-06-26)
Active microbial degraders of the herbicide prosulfocarb (PSC) were isolated to evaluate their performance in soil with a view to their use for bioremediation. The isolated cultures (a microbial consortium and a Pseudomonas sp. strain) were active when tested in
Michèle Nègre et al.
Pest management science, 62(10), 957-964 (2006-08-04)
Adsorption isotherms of prosulfocarb were determined on soils with different physicochemical properties. The extent of adsorption increased with the organic carbon content, but the results also suggested the involvement of some inorganic soil surfaces. In order better to understand the
Roberto Busi et al.
Pest management science, 76(12), 3926-3934 (2020-07-09)
Resistance to the dinitroaniline herbicide trifluralin in Lolium rigidum (annual ryegrass) often is mediated by the enhanced capacity to metabolize the herbicide to less toxic polar conjugates and/or by functionally recessive target-site mutations in α-tubulin. In two L. rigidum populations
Carlos García-Delgado et al.
Journal of hazardous materials, 390, 122162-122162 (2020-02-01)
The spread of organic pollutants from soil to other environments is one important source of environmental pollution. The addition of organic amendments to soil is an interesting strategy to control pollutants leaching. However, the contribution of different carbon types of
Enhanced soil biodegradation of prosulfocarb herbicide in barley crop.
J Rouchaud et al.
Bulletin of environmental contamination and toxicology, 58(5), 752-757 (1997-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service