Skip to Content
Merck
All Photos(3)

Key Documents

296856

Sigma-Aldrich

Diisopropyl ether

anhydrous, 99%, contains either BHT or hydroquinone as stabilizer

Synonym(s):

Isopropyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOCH(CH3)2
CAS Number:
Molecular Weight:
102.17
Beilstein:
1731256
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
Assay:
99%
bp:
68-69 °C (lit.)
vapor pressure:
120 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

vapor density

3.5 (vs air)

vapor pressure

120 mmHg ( 20 °C)

Assay

99%

form

liquid

autoignition temp.

827 °F

contains

stabilizer

expl. lim.

1-21 %, 100 °F

impurities

<0.002% water
<0.005% water (100 mL pkg)

refractive index

n20/D 1.367 (lit.)

bp

68-69 °C (lit.)

mp

−85 °C (lit.)

density

0.725 g/mL at 25 °C (lit.)

SMILES string

CC(C)OC(C)C

InChI

1S/C6H14O/c1-5(2)7-6(3)4/h5-6H,1-4H3

InChI key

ZAFNJMIOTHYJRJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diisopropyl ether may be used as a solvent for the recrystallization of compounds such as N-hydroxysuccinimide esters of acylamino acids[1], 2-acetyl-1,3-cyclopentanedione[2] and cis-1,2-cyclohexanediol.[3]

Other Notes

The article number 296856-6X1L will be discontinued. Please order the single bottle 296856-1L which is physically identical with the same exact specifications.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-20.2 °F - closed cup

Flash Point(C)

-29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Smart morpholine-functional statistical copolymers synthesized by nitroxide mediated polymerization
Lessard BH, et al.
Polymer, 53(25), 5649-5656 (2012)
Catalytic osmium tetroxide oxidation of olefins: cis-1,2-cyclohexanediol
VanRheenen V, et al.
Organic Syntheses, 58, 43-43 (1978)
Atmospheric chemistry of automotive fuel additives: diisopropyl ether.
Wallington TJ, et al.
Environmental Science & Technology, 27(1), 98-104 (1993)
2-Acetyl-1,3-cyclopentanedione
Merenyi F and Nilsson M
Organic Syntheses, 52, 1-1 (1972)
Single stage synthesis of diisopropyl ether-an alternative octane enhancer for lead-free petrol.
Heese FP, et al.
Catalysis Today, 49(1), 327-335 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service