Skip to Content
Merck
All Photos(1)

Key Documents

258350

Sigma-Aldrich

Mesoporphyrin IX dimethyl ester

synthetic

Synonym(s):

Dimethyl 7,12-diethyl-3,8,13,17-tetramethyl-21H,23H-porphine-2,18-dipropionate, NSC 16668

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H42N4O4
CAS Number:
Molecular Weight:
594.74
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

form

solid

reaction suitability

core: porphyrin
reagent type: catalyst

mp

214-215 °C (lit.)

λmax

400 nm

SMILES string

CCc1c(C)c2cc3nc(cc4[nH]c(cc5[nH]c(cc1n2)c(C)c5CC)c(C)c4CCC(=O)OC)c(CCC(=O)OC)c3C

InChI

1S/C36H42N4O4/c1-9-23-19(3)27-15-28-21(5)25(11-13-35(41)43-7)33(39-28)18-34-26(12-14-36(42)44-8)22(6)30(40-34)17-32-24(10-2)20(4)29(38-32)16-31(23)37-27/h15-18,38,40H,9-14H2,1-8H3/b27-15-,28-15-,29-16-,30-17-,31-16-,32-17-,33-18-,34-18-

InChI key

SQMOJNBMZGPIKC-MFBGAUBSSA-N

Application

Reactant for:
  • Metalation reactions
  • H2-hydrogenations
Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service