Skip to Content
Merck
All Photos(3)

Key Documents

213519

Sigma-Aldrich

Pyronin Y

certified by the Biological Stain Commission, Dye content 50 %

Synonym(s):

Pyronin G, Pyronin J

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19ClN2O
CAS Number:
Molecular Weight:
302.80
Colour Index Number:
45005
Beilstein:
3795789
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

certified by the Biological Stain Commission

Quality Level

form

powder or crystals

composition

Dye content, 50%

mp

250-260 °C (lit.)

λmax

548 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].CN(C)c1ccc2C=C3C=C\C(C=C3Oc2c1)=[N+](/C)C

InChI

1S/C17H19N2O.ClH/c1-18(2)14-7-5-12-9-13-6-8-15(19(3)4)11-17(13)20-16(12)10-14;/h5-11H,1-4H3;1H/q+1;/p-1

InChI key

INCIMLINXXICKS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Pyronin Y (C.I. 45005) is an aminoxanthene dye that is present as a weakly lipophilic cation in neutral aqueous solutions. It is also known as pyronine G.

Application

Pyronin Y is commonly used in histology as a constituent of the ethyl (methyl) green-pyronine method for demonstrating DNA and RNA. It is known as a nucleic acid stain for both RNA and DNA. Additionally, it is suitable for other microscopic applications including demonstration of lipofuscin in neurons and simultaneous fixation and staining of phospholipids.

Suitability

Certified for use in the methyl green-pyronin method for differential staining of nucleic acids.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

IL-7 is a potent and proviral strain-specific inducer of latent HIV-1 cellular reservoirs of infected individuals on virally suppressive HAART.
Wang FX
The Journal of Clinical Investigation, 115, 128-137 (2005)
Side population cells contribute to the genesis of human endometrium
Tsuji S
Fertility and Sterility, 90, 1528- 1537 (2008)
Yang Mei et al.
Nature communications, 11(1), 3172-3172 (2020-06-25)
Bone marrow engraftment of the hematopoietic stem and progenitor cells (HSPCs) involves homing to the vasculatures and lodgment to their niches. How HSPCs transmigrate from the vasculature to the niches is unclear. Here, we show that loss of diaphanous-related formin
Pedro Montes-Navajas et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 8(12), 1743-1747 (2009-12-22)
Cucurbiturils (CBs) of the appropriate size (CB[7] and CB[8]) form strong guest-host complexes in phosphate buffer solution (PBS) with acridine orange (AO) and pyronine Y (PYY) with 1 : 1 and 2 : 1 stoichiometries for CB[7] and CB[8] complexes
Yuichiro Koide et al.
ACS chemical biology, 6(6), 600-608 (2011-03-08)
The absorption and emission wavelengths of group 14 pyronines and rhodamines, which contain silicon, germanium, or tin at the 10 position of the xanthene chromophore, showed large bathochromic shifts compared to the original rhodamines, owing to stabilization of the LUMO

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service