Skip to Content
Merck
All Photos(3)

Key Documents

B7920

Sigma-Aldrich

Brilliant Blue R

≥70% purity (HPLC), powder

Synonym(s):

Acid Blue 83, Brilliant indocyanin 6B, Coomassie Brilliant Blue R

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H44N3NaO7S2
CAS Number:
Molecular Weight:
825.97
Colour Index Number:
42660
Beilstein:
5718025
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Brilliant Blue R, pure

Quality Level

Assay

≥70% (HPLC)

form

powder

solubility

H2O: 1 mg/mL

λmax

586-592 nm in ethanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].CCOc1ccc(Nc2ccc(cc2)C(\c3ccc(cc3)N(CC)Cc4cccc(c4)S([O-])(=O)=O)=C5\C=C/C(C=C5)=[N+](\CC)Cc6cccc(c6)S([O-])(=O)=O)cc1

InChI

1S/C45H45N3O7S2.Na/c1-4-47(31-33-9-7-11-43(29-33)56(49,50)51)40-23-15-36(16-24-40)45(35-13-19-38(20-14-35)46-39-21-27-42(28-22-39)55-6-3)37-17-25-41(26-18-37)48(5-2)32-34-10-8-12-44(30-34)57(52,53)54;/h7-30H,4-6,31-32H2,1-3H3,(H2,49,50,51,52,53,54);/q;+1/p-1

InChI key

NKLPQNGYXWVELD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Brilliant Blue R has been used in staining of gels after electrophoresis. It has also been used to determine protein in the protein bodies.
For detection of protein bands following electrophoresis.

Biochem/physiol Actions

Brilliant Blue R is a methanol based stain. It can also stain agarose gels. It consists of methanol and acetic acid.

Legal Information

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Chieh Chang et al.
Scientific reports, 10(1), 11674-11674 (2020-07-17)
Stem cell activity and cell differentiation is robustly influenced by the nutrient availability in the gonads. The signal that connects nutrient availability to gonadal stem cell activity remains largely unknown. In this study, we show that tumor necrosis factor Eiger
Seed structure and localization of reserves in Chenopodium quinoa
Prego, Imelda and Maldonado, Sara and Otegui, Marisa
Annals of Botany, 82(4), 481-488 (1998)
Tenderization effect of soy sauce on beef M. biceps femoris.
Kim HW, et al.
Food Chemistry, 139, 597-597 (2013)
Dong-Heon Song et al.
Foods (Basel, Switzerland), 9(6) (2020-06-06)
The objective of this study is to evaluate the effects of NaCl and KCl salting on technological properties of pre- and post-rigor chicken breasts at various ionic strengths. The following factorial arrangement was used: 2 salt types (NaCl and KCl)
THE
Protein Analysis and Purification: Benchtop Techniques (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service