Skip to Content
Merck
All Photos(2)

Key Documents

213268

Sigma-Aldrich

Calcium hydride

greener alternative

powder, 0-2 mm, reagent grade, ≥90% (gas-volumetric)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2Ca
CAS Number:
Molecular Weight:
42.09
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥90% (gas-volumetric)
grade:
reagent grade
form:
powder

grade

reagent grade

Quality Level

Assay

≥90% (gas-volumetric)

form

powder

reaction suitability

reagent type: reductant

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

particle size

0-2 mm

mp

816 °C (lit.)

greener alternative category

SMILES string

[Ca]

InChI

1S/Ca.2H

InChI key

FAQLAUHZSGTTLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Calcium hydride is an alkaline earth metal hydride. Its solid state reaction with hexachlorobenzene has been investigated.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Calcium hydride (CaH2) may be employed as a reducing agent in the one-pot two-step Knoevenagel condensation reaction.
It may also be used as a reagent furing metallothermic synthesis of Nb3Al.
CaH2 can be used as a hydrogen source during the synthesis of monomethylated and dimethylated amines and in a ball milling process for the dechlorination of liquid and solid chlorinated compounds.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modification of Calcium Hydride as Solid Hydrogen Source for Fuel Cell System.
Nagashima K, et al.
International Journal of Applied Ceramic Technology, 13(2), 265-268 (2016)
Reductive N-methylation of amines with calcium hydride and Pd/C catalyst.
Guyon C, et al.
Tetrahedron Letters (2016)
Properties of the intermetallic phase Nb3Al prepared by a calcium hydride reduction process.
Kasimtsev AV, et al.
Inorganic Materials, 51(1), 43-50 (2015)
L R Cajero-Zul et al.
Journal of biomaterials science. Polymer edition, 30(3), 163-189 (2018-12-18)
Nanocomposites of functionalized carbon nanotubes (CNTsf) as nanofillers, and a copolymer of star-shaped poly(ε-caprolactone) (stPCL) and poly(ethylene glycol) (PEG) as a polymeric matrix were synthesized, characterized, and their resistance to the growth of Staphylococcus aureus and Pseudomonas aeruginosa was evaluated.
Selective mechanochemical dehalogenation of chlorobenzenes over calcium hydride.
Loiselle S, et al.
Environmental Science & Technology, 31(1, 261-265 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service