Skip to Content
Merck
All Photos(1)

Key Documents

109606

Sigma-Aldrich

Methacrylamide

98%

Synonym(s):

2-Methylacrylamide, 2-Methylpropenamide, Methacrylic acid amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CONH2
CAS Number:
Molecular Weight:
85.10
Beilstein:
605397
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

solid

mp

106-110 °C (lit.)

SMILES string

CC(=C)C(N)=O

InChI

1S/C4H7NO/c1-3(2)4(5)6/h1H2,2H3,(H2,5,6)

InChI key

FQPSGWSUVKBHSU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methacrylamide is a functional monomer used inthe preparation of thermo-sensitive hydrogels, polymer latexes, and self-etchingprimers with a longer shelf life. It can also be used to prepare dispersionsuspensions to synthesize biocompatible HA/sol-gel glass mixtures.

Application

  • Synthesis of non-fouling poly brushes by photoinduced SET-LRP: This study highlights the use of photoinduced SET-LRP for the polymerization of methacrylamide, emphasizing its efficiency and the quality of the resulting polymers for non-fouling applications (M Vorobii, A de los Santos Pereira, 2015).
  • Hydrolytic stability of methacrylamide and methacrylate in gelatin methacryloyl: The study investigates the hydrolytic stability of methacrylamide within gelatin methacryloyl, highlighting its stability and potential in biomedical applications (J Zheng, M Zhu, G Ferracci, NJ Cho, 2018).
  • Two-step mechanisms of tumor selective delivery of N-(2-hydroxypropyl) methacrylamide copolymer conjugated with pirarubicin via an acid-cleavable linkage: This paper discusses the development of a copolymer conjugate for targeted cancer therapy, showcasing a two-step mechanism for enhanced drug delivery (H Nakamura, T Etrych, P Chytil, M Ohkubo, 2014).
  • Backbone Degradable N-(2-Hydroxypropyl)methacrylamide Copolymer Conjugates with Gemcitabine and Paclitaxel: The research focuses on degradable copolymer conjugates for delivering cancer therapeutics, noting significant effects on tumor reduction and highlighting the impact of molecular weight (J Yang, R Zhang, H Pan, Y Li, Y Fang, 2017).

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT RE 2 - STOT SE 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sinoj Abraham et al.
Biomacromolecules, 12(10), 3567-3580 (2011-09-07)
Nonfouling polymer architectures are considered important to the successful implementation of many biomaterials. It is thought that how these polymers induce conformational changes in proteins upon adsorption may dictate the fate of the device being utilized. Herein, oxidized silicon nanoparticles
Hoyong Chung et al.
Biomacromolecules, 12(2), 342-347 (2010-12-25)
We present a study on the effects of cross-linking on the adhesive properties of bio-inspired 3,4-dihydroxyphenylalanine (DOPA). DOPA has a unique catechol moiety found in adhesive proteins in marine organisms, such as mussels and polychaete, which results in strong adhesion
Patrik Sobolčiak et al.
Macromolecular rapid communications, 34(8), 635-639 (2013-02-13)
A novel cationic polymer poly(N,N-dimethyl-N-[3-(methacroylamino) propyl]-N-[2-[(2-nitrophenyl)methoxy]-2-oxo-ethyl]ammonium chloride) is synthesized by free-radical polymerization of N-[3-(dimethylamino)propyl] methacrylamide and subsequent quaternization with o-nitrobenzyl 2-chloroacetate. The photolabile o-nitrobenzyl carboxymethyl pendant moiety is transformed to the zwitterionic carboxybetaine form upon the irradiation at 365 nm.
Cesar Rodriguez-Emmenegger et al.
Macromolecular rapid communications, 32(13), 958-965 (2011-06-08)
Among the class of zwitterionic polymers poly(carboxybetaine)s (poly(CB)s) are unique, emerging as the only ultra-low fouling materials known allowing the preparation of biosensors, fouling resistant nanoparticles, and non-adhesive surfaces for bacteria. Poly(carboxybetaine methacrylate) and poly(carboxybetaine acrylamide) have been prepared via
Ellen Verheyen et al.
Journal of controlled release : official journal of the Controlled Release Society, 156(3), 329-336 (2011-09-14)
We report an efficient strategy to conjugate methacrylamide moieties to the lysine units of lysozyme for co-polymerization and subsequent triggered release from hydrogels. Two novel linker molecules, containing an ester bond and/or a disulfide bond for temporary immobilization, were synthesized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service