Skip to Content
Merck
All Photos(3)

Key Documents

201049

Sigma-Aldrich

Lithium hydride

greener alternative

powder, −30 mesh, ≥95%

Synonym(s):

LiH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiH
CAS Number:
Molecular Weight:
7.95
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥95%

form

powder

reaction suitability

reagent type: reductant

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

particle size

−30 mesh

mp

680 °C (lit.)

density

0.82 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

[Li]

InChI

1S/Li.H

InChI key

SIAPCJWMELPYOE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium hydride is a simple neutral heteropolar diatomic molecule. It liberates hydrogen upon heating it to temperatures above 500oC. Lithium hydride, like lithium aluminum hydride, exhibits a high gravimetric hydrogen storage capacity. LiH is known to have large specific heat and latent heat of fusion.5
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Lithium hydride may be used as an effective neutron shield for mobile nuclear reactors. LiH may also be used as a rocket fuel, energy storage system, and heat shield.5

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Thermophysical Properties of Chemicals and Hydrocarbons null
Spectroscopy and structure of the lithium hydride diatomic molecules and ions.
Stwalley WC and Zemke WT
The Journal of Physical Chemistry, 22(1), 87-112 (1993)
Lithium hydride: A space age shielding material
Welch FH
Nuclear Engineering and Design, 26(3), 444-460 (1974)
Kiyotaka Goshome et al.
Molecules (Basel, Switzerland), 24(7) (2019-04-10)
Hydrogenation of a lithium-potassium (double-cation) amide (LiK(NH₂)₂), which is generated as a product by ammonolysis of litium hydride and potassium hydride (LiH-KH) composite, is investigated in details. As a result, lithium amide (LiNH₂) and KH are generated after hydrogenation at
Josué A Torres-Ávalos et al.
Polymers, 13(4) (2021-03-07)
Design of a smart drug delivery system is a topic of current interest. Under this perspective, polymer nanocomposites (PNs) of butyl acrylate (BA), methacrylic acid (MAA), and functionalized carbon nanotubes (CNTsf) were synthesized by in situ emulsion polymerization (IEP). Carbon

Articles

Mechanochemical Effect of Severe Plastic Deformations: Metal Alloys, Hydrides and Molecular Solids

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service