Skip to Content
Merck
All Photos(1)

Key Documents

11709

Supelco

Barbituric acid

for spectrophotometric det. of cyanide, ≥99.5%

Synonym(s):

2,4,6-Trihydroxypyrimidine, Malonylurea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N2O3
CAS Number:
Molecular Weight:
128.09
Beilstein:
120502
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥99.5% (HPLC)
≥99.5%

form

solid

quality

for spectrophotometric det. of cyanide

technique(s)

UV/Vis spectroscopy: suitable

ign. residue

≤0.05%

mp

248-252 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤500 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Mg: ≤10 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

O=C1CC(=O)NC(=O)N1

InChI

1S/C4H4N2O3/c7-2-1-3(8)6-4(9)5-2/h1H2,(H2,5,6,7,8,9)

InChI key

HNYOPLTXPVRDBG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Barbituric acid is also known as malonurea or 6-hydroxyuracil, it is an odourless powder and is soluble in water. It has a very high pharmacological activity and can be used to synthesize addition derivatives of it, to be used in novel drug discovery. It basically acts on the central nervous system (CNS) depressants, thereby possessing a wide range from mild sedation to total anaesthesia.

Application

Barbituric acid with aromatic aldehydes was used in an experimental study, meant to demonstrate the increased efficiency of Knoevenagel condensation reaction for barbituric acid and various aromatic aldehydes on basic alumina, in the absence of organic solvents under microwave irradiation. It may also be used in electrochemical oxidation of iodine, using cyclic voltammetry and controlled-potential coulometry.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bioactive Heterocyclic Compound Classes.
Clemens L and Dinges J.
Pharmaceutics, 24-27 (2012)
Microwave enhanced knoevenagel condensation of barbituric acid with aromatic aldehydes on basic alumina.
Khalafi NA and Hashemi A.
Iranian Journal of Chemistry and Chemical Engineering, 20 (1), 9-11 (2001)
Electrochemical study of iodide in the presence of barbituric acid. Application to coulometric titration of barbituric acid.
Nematollahi D and Hesari M
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 70 (1), 7-11 (2001)
David C Chiara et al.
The Journal of biological chemistry, 288(27), 19343-19357 (2013-05-17)
GABA type A receptors (GABAAR), the brain's major inhibitory neurotransmitter receptors, are the targets for many general anesthetics, including volatile anesthetics, etomidate, propofol, and barbiturates. How such structurally diverse agents can act similarly as positive allosteric modulators of GABAARs remains
Sergio Hidalgo-Figueroa et al.
Chemical biology & drug design, 81(4), 474-483 (2013-01-08)
A small series of thiazolidine-2,4-dione and barbituric acid derivatives 1-4 was prepared using a short synthetic route, and all compounds were characterized by elemental analysis, mass spectrometry, and NMR ((1)H, (13)C) spectroscopy. Their in vitro relative expression of peroxisome proliferator-activated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service