Skip to Content
Merck
All Photos(1)

Key Documents

1.19533

Supelco

Cyanide standard solution

traceable to SRM from NIST K₂[Zn(CN)₄] in H₂O 1000 mg/l CN Certipur®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

Quality Level

technique(s)

ion chromatography: suitable

pH

9-10 (20 °C in H2O)

density

1.00 g/cm3 at 20 °C

application(s)

environmental
food and beverages

format

single component solution

storage temp.

2-8°C

Application

  • Development of Cyanide Detection Methods: Utilizing high-performance liquid chromatography-tandem mass spectrometry, researchers have developed concurrent determination methods for cyanide and thiocyanate in human and swine blood. This technique ensures high accuracy and sensitivity, crucial for toxicological analysis and forensic investigations (Alluhayb et al., 2023).
  • Fluorometric Cyanide Sensing: A novel fluorometric probe offers high selectivity for sensing cyanide in aqueous solutions, which can be applied to quantitative determinations and living cell imaging. This advancement supports biochemical assays requiring precise cyanide measurements, enhancing safety and detection capabilities in both environmental and clinical settings (Satheeshkumar et al., 2023).

Analysis Note

Concentration     β (CN⁻): 990 - 1010 mg/l
Determination method: Argentometric titration.
The content of this solution was determined with silver nitrate standard solution (article number 1.09081) standardized against volumetric standard sodium chloride (article number 1.02406). The expanded measurement uncertainty is ± 0.7 % (k=2 coverage factor for 95% coverage probability). The certified value is traceable to primary standard NIST SRM 999b (NIST: National Institute of Standards and Technology, USA) by means of volumetric standard sodium chloride, measured in the accredited calibration laboratory of Merck KGaA, Darmstadt, Germany in accordance to DIN EN ISO/IEC 17025.

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Supplementary Hazards

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service